missing translation for 'onlineSavingsMsg'
Learn More

3-chloro-2-methylbenzenesulfonyl chloride, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 357080010

 View more versions of this product

Catalog No. AC357080010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

3-chloro-2-methylbenzenesulfonyl chloride
97.5
Authentic
Glass bottle
Cl(CH3)C6H3SO2Cl
1g
ZSIYKAQPQRTBPF-UHFFFAOYSA-N
3-chloro-2-methylbenzenesulfonyl chloride
522734
98%
80563-86-6
100.0
97.5% min. (ex Cl) (Argentometry)
C7H6Cl2O2S
MFCD00052198
3-chloro-2-methylbenzene-1-sulfonyl chloride, 3-chloro-2-methylbenzenesulphonyl chloride, benzenesulfonyl chloride, 3-chloro-2-methyl-9ci, 3-chloro-2-methylphenylsulfonyl chloride, 3-chloro-2-methyl-benzenesulfonyl chloride, chloro 3-chloro-2-methylphenyl sulfone, pubchem5096, acmc-209pjq, buttpark 3711-61, 3-chloro-2-methylbenzenesulfonylchloride
CC1=C(C=CC=C1Cl)S(=O)(=O)Cl
225.09
225.09

Chemical Identifiers

80563-86-6
225.09
ZSIYKAQPQRTBPF-UHFFFAOYSA-N
522734
CC1=C(C=CC=C1Cl)S(=O)(=O)Cl
C7H6Cl2O2S
MFCD00052198
3-chloro-2-methylbenzene-1-sulfonyl chloride, 3-chloro-2-methylbenzenesulphonyl chloride, benzenesulfonyl chloride, 3-chloro-2-methyl-9ci, 3-chloro-2-methylphenylsulfonyl chloride, 3-chloro-2-methyl-benzenesulfonyl chloride, chloro 3-chloro-2-methylphenyl sulfone, pubchem5096, acmc-209pjq, buttpark 3711-61, 3-chloro-2-methylbenzenesulfonylchloride
3-chloro-2-methylbenzenesulfonyl chloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only