missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Chloro-2-methylpropene, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B226740B

 View more versions of this product

Catalog No. AAB226740B


Explore available promotions

Description

Description

3-Chloro-2-methylpropene has been used as reactant in the synthesis of cyclobutanone. It is used as an intermediate in the production of plastics, pharmaceuticals and other organic chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Chloro-2-methylpropene has been used as reactant in the synthesis of cyclobutanone. It is used as an intermediate in the production of plastics, pharmaceuticals and other organic chemicals.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

3-Chloro-2-methylpropene
-80°C
71°C to 72°C
C4H7Cl
MFCD00000953
UN2554
3-chloro-2-methylpropene, methallyl chloride, 3-chloro-2-methyl-1-propene, 1-propene, 3-chloro-2-methyl, 2-methylallyl chloride, isobutenyl chloride, 2-methyl-2-propenyl chloride, beta-methallyl chloride, propene, 3-chloro-2-methyl, cloruro di metallile
OHXAOPZTJOUYKM-UHFFFAOYSA-N
3-chloro-2-methylprop-1-ene
11241
90.55
563-47-3
0.917
−12°C (9°F)
1.4275
1000 g
14,2148
Not miscible in water.
CC(=C)CCl
90.55
CHEBI:82419
98%

Chemical Identifiers

563-47-3
90.55
OHXAOPZTJOUYKM-UHFFFAOYSA-N
11241
3-chloro-2-methylprop-1-ene
C4H7Cl
MFCD00000953
3-chloro-2-methylpropene, methallyl chloride, 3-chloro-2-methyl-1-propene, 1-propene, 3-chloro-2-methyl, 2-methylallyl chloride, isobutenyl chloride, 2-methyl-2-propenyl chloride, beta-methallyl chloride, propene, 3-chloro-2-methyl, cloruro di metallile
CHEBI:82419
CC(=C)CCl
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H314-H302-H332-H317
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful if inhaled.
May cause an allergic skin reaction.

P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P272-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P372-P374-P380-P501c

H225-H302+H332-H314-H317-H335-H350

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: METHYL ALLYL CHLORIDE

EINECSNumber : 209-251-2

RTECSNumber : UC8050000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only