missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Chloro-2-nitrotoluene 96.0+%, TCI America™
Supplier: TCI America C27315G
Specifications
3-Chloro-2-nitrotoluene | |
Yellow | |
C15H22N2O4 | |
5 g | |
QTKPBWWPUDOQGJ-UHFFFAOYSA-N | |
4-[5-(4-nitrophenoxy)pentyl]morpholine | |
79328 | |
≥96.0% (GC) |
5367-26-0 | |
80°C | |
MFCD04117977 | |
2433 | |
[O-][N+](=O)C1=CC=C(OCCCCCN2CCOCC2)C=C1 | |
294.35 | |
171.58 | |
Liquid |
Chemical Identifiers
5367-26-0 | |
294.35 | |
QTKPBWWPUDOQGJ-UHFFFAOYSA-N | |
4-[5-(4-nitrophenoxy)pentyl]morpholine |
C15H22N2O4 | |
MFCD04117977 | |
79328 | |
[O-][N+](=O)C1=CC=C(OCCCCCN2CCOCC2)C=C1 |
Safety and Handling
EINECSNumber : (3)-0449
TSCA : No