missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Chloro-3-methyl-1-butyne, 95%, Thermo Scientific Chemicals

$242.88 - $810.82

Chemical Identifiers

CAS 1111-97-3
Molecular Formula C5H7Cl
Molecular Weight (g/mol) 102.56
MDL Number MFCD00190221
InChI Key QSILYWCNPOLKPN-UHFFFAOYSA-N
Synonym 3-chloro-3-methyl-1-butyne, 1-butyne, 3-chloro-3-methyl, 3-chloro-3-methylbutyne, 3-methyl-3-chlorobutyne, acmc-1bo5c, 2-chloro-2-methyl butyne, 3-chloro-3-methyl butyne, 3-chloro-3-methyl-butyne, 3-chloro-3-methyl-1-butyn, 2-chloro-2-methyl-3-butyne
PubChem CID 70679
IUPAC Name 3-chloro-3-methylbut-1-yne
SMILES CC(C)(Cl)C#C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2525206
View Documents
Thermo Scientific Chemicals
B2525206
5 g
Each for $242.88
 
AAB2525214
View Documents
Thermo Scientific Chemicals
B2525214
25 g
Each for $810.82
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1111-97-3
102.56
QSILYWCNPOLKPN-UHFFFAOYSA-N
70679
CC(C)(Cl)C#C
C5H7Cl
MFCD00190221
3-chloro-3-methyl-1-butyne, 1-butyne, 3-chloro-3-methyl, 3-chloro-3-methylbutyne, 3-methyl-3-chlorobutyne, acmc-1bo5c, 2-chloro-2-methyl butyne, 3-chloro-3-methyl butyne, 3-chloro-3-methyl-butyne, 3-chloro-3-methyl-1-butyn, 2-chloro-2-methyl-3-butyne
3-chloro-3-methylbut-1-yne

Specifications

1111-97-3
0.913
4°C (40°F)
C5H7Cl
MFCD00190221
UN1993
QSILYWCNPOLKPN-UHFFFAOYSA-N
3-chloro-3-methylbut-1-yne
70679
95%
-61°C
73°C to 75°C
Recognizable
1.4178
5 g
3-chloro-3-methyl-1-butyne, 1-butyne, 3-chloro-3-methyl, 3-chloro-3-methylbutyne, 3-methyl-3-chlorobutyne, acmc-1bo5c, 2-chloro-2-methyl butyne, 3-chloro-3-methyl butyne, 3-chloro-3-methyl-butyne, 3-chloro-3-methyl-1-butyn, 2-chloro-2-methyl-3-butyne
CC(C)(Cl)C#C
102.56
102.57
3-Chloro-3-methyl-1-butyne
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c

H225-H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

EINECSNumber : 214-188-9

TSCA : Yes

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only