missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Chloro-4-fluorobenzeneboronic acid, 98%, Thermo Scientific Chemicals

$159.79 - $600.81

Chemical Identifiers

CAS 144432-85-9
Molecular Formula C6H5BClFO2
Molecular Weight (g/mol) 174.362
MDL Number MFCD00051800
InChI Key WJDZZXIDQYKVDG-UHFFFAOYSA-N
Synonym 3-chloro-4-fluorobenzeneboronic acid, 3-chloro-4-fluorophenyl boronic acid, 3-chloro-4-fluorophenyl boranediol, 4-fluoro-3-chlorophenylboronic acid, 3-chloro-4-fluoro phenyl boronic acid, 3-chloro-4-fluoro-phenyl boronic acid, boronic acid, 3-chloro-4-fluorophenyl, b-3-chloro-4-fluorophenyl-boronic acid, pubchem1780
PubChem CID 2734660
IUPAC Name (3-chloro-4-fluorophenyl)boronic acid
SMILES B(C1=CC(=C(C=C1)F)Cl)(O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2275506
View Documents
Thermo Scientific Chemicals
B2275506
5 g
Each for $159.79
 
AAB2275514
View Documents
Thermo Scientific Chemicals
B2275514
25 g
Each for $600.81
 
Description

Description

3-Chloro-4-fluorobenzeneboronic acid is used as a reactant for Rh-catalyzed asymmetric addition reactions, palladium-catalyzed oxidative cross-coupling reaction and Suzuki-Miyaura coupling.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Chloro-4-fluorobenzeneboronic acid is used as a reactant for Rh-catalyzed asymmetric addition reactions, palladium-catalyzed oxidative cross-coupling reaction and Suzuki-Miyaura coupling.

Notes
Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

144432-85-9
174.362
WJDZZXIDQYKVDG-UHFFFAOYSA-N
2734660
B(C1=CC(=C(C=C1)F)Cl)(O)O
C6H5BClFO2
MFCD00051800
3-chloro-4-fluorobenzeneboronic acid, 3-chloro-4-fluorophenyl boronic acid, 3-chloro-4-fluorophenyl boranediol, 4-fluoro-3-chlorophenylboronic acid, 3-chloro-4-fluoro phenyl boronic acid, 3-chloro-4-fluoro-phenyl boronic acid, boronic acid, 3-chloro-4-fluorophenyl, b-3-chloro-4-fluorophenyl-boronic acid, pubchem1780
(3-chloro-4-fluorophenyl)boronic acid

Specifications

144432-85-9
C6H5BClFO2
5 g
3-chloro-4-fluorobenzeneboronic acid, 3-chloro-4-fluorophenyl boronic acid, 3-chloro-4-fluorophenyl boranediol, 4-fluoro-3-chlorophenylboronic acid, 3-chloro-4-fluoro phenyl boronic acid, 3-chloro-4-fluoro-phenyl boronic acid, boronic acid, 3-chloro-4-fluorophenyl, b-3-chloro-4-fluorophenyl-boronic acid, pubchem1780
B(C1=CC(=C(C=C1)F)Cl)(O)O
174.362
174.37
3-Chloro-4-fluorobenzeneboronic acid
255°C to 260°C
MFCD00051800
7369789
WJDZZXIDQYKVDG-UHFFFAOYSA-N
(3-chloro-4-fluorophenyl)boronic acid
2734660
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only