missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-fluorobenzotrifluoride, 98%

CAS: 78068-85-6 | C7H3ClF4 | 198.54 g/mol

$164.34 - $669.05

Chemical Identifiers

CAS 78068-85-6
Molecular Formula C7H3ClF4
Molecular Weight (g/mol) 198.54
MDL Number MFCD00042207
InChI Key BKHVEYHSOXVAOP-UHFFFAOYSA-N
Synonym 3-chloro-4-fluorobenzotrifluoride, 2-chloro-1-fluoro-4-trifluoromethyl benzene, benzene, 2-chloro-1-fluoro-4-trifluoromethyl, 3-cholor-4-fluorobenzo-trifluoride, 3-chloro-4-fluoro benzotrifluoride, 1-chloro-2-fluoro-5-trifluoromethyl benzene, 3-chloro-4-fluoro-1-trifluoromethyl benzene, benzene, chloro-1-fluoro-4-trifluoromethyl, 3-chloro-alpha,alpha,alpha,4-tetrafluorotoluene, 3-chloro-4-fluoro-alpha,alpha,alpha-trifluorotoluene
PubChem CID 157153
IUPAC Name 2-chloro-1-fluoro-4-(trifluoromethyl)benzene
SMILES FC1=CC=C(C=C1Cl)C(F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1667006
View Documents
Thermo Scientific Chemicals
A1667006
5 g
Each for $164.34
Only null left
 
AAA1667014
View Documents
Thermo Scientific Chemicals
A1667014
25 g
Each for $669.05
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

78068-85-6
198.54
BKHVEYHSOXVAOP-UHFFFAOYSA-N
157153
FC1=CC=C(C=C1Cl)C(F)(F)F
C7H3ClF4
MFCD00042207
3-chloro-4-fluorobenzotrifluoride, 2-chloro-1-fluoro-4-trifluoromethyl benzene, benzene, 2-chloro-1-fluoro-4-trifluoromethyl, 3-cholor-4-fluorobenzo-trifluoride, 3-chloro-4-fluoro benzotrifluoride, 1-chloro-2-fluoro-5-trifluoromethyl benzene, 3-chloro-4-fluoro-1-trifluoromethyl benzene, benzene, chloro-1-fluoro-4-trifluoromethyl, 3-chloro-alpha,alpha,alpha,4-tetrafluorotoluene, 3-chloro-4-fluoro-alpha,alpha,alpha-trifluorotoluene
2-chloro-1-fluoro-4-(trifluoromethyl)benzene

Specifications

78068-85-6
137°C
C7H3ClF4
MFCD00042207
UN1993
3-chloro-4-fluorobenzotrifluoride, 2-chloro-1-fluoro-4-trifluoromethyl benzene, benzene, 2-chloro-1-fluoro-4-trifluoromethyl, 3-cholor-4-fluorobenzo-trifluoride, 3-chloro-4-fluoro benzotrifluoride, 1-chloro-2-fluoro-5-trifluoromethyl benzene, 3-chloro-4-fluoro-1-trifluoromethyl benzene, benzene, chloro-1-fluoro-4-trifluoromethyl, 3-chloro-alpha,alpha,alpha,4-tetrafluorotoluene, 3-chloro-4-fluoro-alpha,alpha,alpha-trifluorotoluene
FC1=CC=C(C=C1Cl)C(F)(F)F
198.54
198.55
3-Chloro-4-fluorobenzotrifluoride
1.49
42°C (108°F)
1.4355
5 g
4309386
BKHVEYHSOXVAOP-UHFFFAOYSA-N
2-chloro-1-fluoro-4-(trifluoromethyl)benzene
157153
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H226-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only