missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Chloro-4-fluorophenol, 97%, Thermo Scientific Chemicals

$62.45 - $62.45

Chemical Identifiers

CAS 2613-23-2
Molecular Formula C6H4ClFO
Molecular Weight (g/mol) 146.545
MDL Number MFCD00002257
InChI Key ZQXLIXHVJVAPLW-UHFFFAOYSA-N
Synonym phenol, 3-chloro-4-fluoro, 3-chloro-4-fluoro-phenol, pubchem1492, pubchem2856, 3-chloro-4-fluorophe, 3-chloro4-fluorophenol, acmc-209goi, 4-fluoro-3-chlorophenol, 3-chloro-4-fluorophenol, ksc204s3j
PubChem CID 75790
IUPAC Name 3-chloro-4-fluorophenol
SMILES C1=CC(=C(C=C1O)Cl)F
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1438206
View Documents
Thermo Scientific Chemicals
A1438206
5 g
Each for $62.45
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2613-23-2
146.545
ZQXLIXHVJVAPLW-UHFFFAOYSA-N
75790
C1=CC(=C(C=C1O)Cl)F
C6H4ClFO
MFCD00002257
phenol, 3-chloro-4-fluoro, 3-chloro-4-fluoro-phenol, pubchem1492, pubchem2856, 3-chloro-4-fluorophe, 3-chloro4-fluorophenol, acmc-209goi, 4-fluoro-3-chlorophenol, 3-chloro-4-fluorophenol, ksc204s3j
3-chloro-4-fluorophenol

Specifications

2613-23-2
103°C to 104°C (11 mmHg)
Phenol-like
MFCD00002257
UN3261
phenol, 3-chloro-4-fluoro, 3-chloro-4-fluoro-phenol, pubchem1492, pubchem2856, 3-chloro-4-fluorophe, 3-chloro4-fluorophenol, acmc-209goi, 4-fluoro-3-chlorophenol, 3-chloro-4-fluorophenol, ksc204s3j
C1=CC(=C(C=C1O)Cl)F
146.545
146.55
3-Chloro-4-fluorophenol
38°C to 43°C
108°C (226°F)
C6H4ClFO
5 g
3236443
ZQXLIXHVJVAPLW-UHFFFAOYSA-N
3-chloro-4-fluorophenol
75790
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H302-H312
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 220-042-5

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only