missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Chloro-4-hydroxybenzonitrile, 95%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H5043403

 View more versions of this product

Catalog No. AAH5043403


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-Chloro-4-hydroxybenzonitrile
147°C to 149°C
MFCD01567246
benzonitrile, 3-chloro-4-hydroxy, 2-chloro-4-cyanophenol, 3-chloro-4-hydroxy-benzonitrile, 3-chloro-4-hydroxybenzenecarbonitrile, pubchem4726, 4-cyano-2-chlorophenol, buttpark 4357-94, acmc-1clj1, pharmabridge p-1867, buttpark 43\57-94
C1=CC(=C(C=C1C#N)Cl)O
153.565
153.57
2315-81-3
C7H4ClNO
1 g
CRYPJUOSZDQWJZ-UHFFFAOYSA-N
3-chloro-4-hydroxybenzonitrile
2735739
95%

Chemical Identifiers

2315-81-3
153.565
CRYPJUOSZDQWJZ-UHFFFAOYSA-N
2735739
C1=CC(=C(C=C1C#N)Cl)O
C7H4ClNO
MFCD01567246
benzonitrile, 3-chloro-4-hydroxy, 2-chloro-4-cyanophenol, 3-chloro-4-hydroxy-benzonitrile, 3-chloro-4-hydroxybenzenecarbonitrile, pubchem4726, 4-cyano-2-chlorophenol, buttpark 4357-94, acmc-1clj1, pharmabridge p-1867, buttpark 43\57-94
3-chloro-4-hydroxybenzonitrile
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only