missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3'-Chloro-4'-methoxyacetophenone 98.0+%, TCI America™

Supplier:  TCI America C30931G

 View more versions of this product

Catalog No. C30931G


Explore available promotions

Description

Description

Specifications

Specifications

3′-Chloro-4′-methoxyacetophenone
76°C
148°C
MFCD04973413
3-chloro-4-methoxyacetophenone, 1-3-chloro-4-methoxyphenyl ethanone, 3'-chloro-4'-methoxyacetophenone, 1-3-chloro-4-methoxyphenyl ethan-1-one, ethanone, 1-3-chloro-4-methoxyphenyl, pubchem16367, 3-chloro-4-methoxy acetophenone, 3'-chloro-4'-methoxy acetophenone, 3-cl-4-ch3o-c6h3-coch3
CC(=O)C1=CC(=C(C=C1)OC)Cl
184.619
184.62
Crystalline Powder
37612-52-5
White-Yellow
C9H9ClO2
1 g
QILWOKAXHOAFOF-UHFFFAOYSA-N
1-(3-chloro-4-methoxyphenyl)ethanone
520857
≥98.0% (GC)

Chemical Identifiers

37612-52-5
184.619
QILWOKAXHOAFOF-UHFFFAOYSA-N
520857
CC(=O)C1=CC(=C(C=C1)OC)Cl
C9H9ClO2
MFCD04973413
3-chloro-4-methoxyacetophenone, 1-3-chloro-4-methoxyphenyl ethanone, 3'-chloro-4'-methoxyacetophenone, 1-3-chloro-4-methoxyphenyl ethan-1-one, ethanone, 1-3-chloro-4-methoxyphenyl, pubchem16367, 3-chloro-4-methoxy acetophenone, 3'-chloro-4'-methoxy acetophenone, 3-cl-4-ch3o-c6h3-coch3
1-(3-chloro-4-methoxyphenyl)ethanone
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions