missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Chloro-4-methylaniline 97.0+%, TCI America™

Supplier:  TCI America C0538500G

 View more versions of this product

Catalog No. C0538500G


Explore available promotions

Description

Description

Specifications

Specifications

3-Chloro-4-methylaniline
White-Yellow
C7H8ClN
500 g
3-chloro-p-toluidine, 2-chloro-4-aminotoluene, 4-amino-2-chlorotoluene, benzenamine, 3-chloro-4-methyl, 3-chloro-4-methylbenzenamine, 3-chloro-4-methylphenylamine, 1-amino-3-chloro-4-methylbenzene, 4-methyl-3-chloroaniline, p-toluidine, 3-chloro, gull toxicant
CC1=C(C=C(C=C1)N)Cl
141.598
CHEBI:37824
≥97.0% (GC)
95-74-9
244°C
MFCD00007773
2239
RQKFYFNZSHWXAW-UHFFFAOYSA-N
3-chloro-4-methylaniline
7255
141.60
Crystalline Lumps

Chemical Identifiers

95-74-9
141.598
RQKFYFNZSHWXAW-UHFFFAOYSA-N
7255
3-chloro-4-methylaniline
C7H8ClN
MFCD00007773
3-chloro-p-toluidine, 2-chloro-4-aminotoluene, 4-amino-2-chlorotoluene, benzenamine, 3-chloro-4-methyl, 3-chloro-4-methylbenzenamine, 3-chloro-4-methylphenylamine, 1-amino-3-chloro-4-methylbenzene, 4-methyl-3-chloroaniline, p-toluidine, 3-chloro, gull toxicant
CHEBI:37824
CC1=C(C=C(C=C1)N)Cl
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0265

RTECSNumber : XU5111000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions