missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Chloro-5-hydroxybenzoic Acid 98.0+%, TCI America™
$216.24 - $728.84
Chemical Identifiers
| CAS | 53984-36-4 |
|---|---|
| Molecular Formula | C7H5ClO3 |
| Molecular Weight (g/mol) | 172.56 |
| MDL Number | MFCD04114327 |
| InChI Key | RJOLIYHZZKAIET-UHFFFAOYSA-N |
| PubChem CID | 13071646 |
| IUPAC Name | 3-chloro-5-hydroxybenzoic acid |
| SMILES | OC(=O)C1=CC(O)=CC(Cl)=C1 |
Chemical Identifiers
| 53984-36-4 | |
| 172.56 | |
| RJOLIYHZZKAIET-UHFFFAOYSA-N | |
| 3-chloro-5-hydroxybenzoic acid |
| C7H5ClO3 | |
| MFCD04114327 | |
| 13071646 | |
| OC(=O)C1=CC(O)=CC(Cl)=C1 |
Specifications
| 53984-36-4 | |
| White-Yellow | |
| MFCD04114327 | |
| RJOLIYHZZKAIET-UHFFFAOYSA-N | |
| 3-chloro-5-hydroxybenzoic acid | |
| 13071646 | |
| ≥98.0% (GC,T) | |
| 3-Chloro-5-hydroxybenzoic Acid |
| 246°C | |
| C7H5ClO3 | |
| 1 g | |
| OC(=O)C1=CC(O)=CC(Cl)=C1 | |
| 172.56 | |
| 172.56 | |
| Crystalline Powder |
Safety and Handling
TSCA : No