Learn More
3-Chloro-5-(trifluoromethyl)phenol, 97%, Thermo Scientific™
$143.90 - $143.90
Chemical Identifiers
CAS | 570391-18-3 |
---|---|
Molecular Formula | C7H4ClF3O |
Molecular Weight (g/mol) | 196.55 |
MDL Number | MFCD04115852 |
InChI Key | BPJWMOPEHPOKFT-UHFFFAOYSA-N |
Synonym | 3-chloro-5-trifluoromethyl phenol, 3-chloro-5-hydroxybenzotrifluoride, 3-chloro-5-trifluoromethyl-phenol, 3-trifluoromethyl-5-chlorophenol, 5-chloro-3-trifluoromethyl phenol, phenol, 3-chloro-5-trifluoromethyl |
PubChem CID | 3852718 |
IUPAC Name | 3-chloro-5-(trifluoromethyl)phenol |
SMILES | OC1=CC(Cl)=CC(=C1)C(F)(F)F |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH2632103
|
Thermo Scientific Chemicals
H2632103 |
1 g |
N/A
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
570391-18-3 | |
196.55 | |
BPJWMOPEHPOKFT-UHFFFAOYSA-N | |
3852718 | |
OC1=CC(Cl)=CC(=C1)C(F)(F)F |
C7H4ClF3O | |
MFCD04115852 | |
3-chloro-5-trifluoromethyl phenol, 3-chloro-5-hydroxybenzotrifluoride, 3-chloro-5-trifluoromethyl-phenol, 3-trifluoromethyl-5-chlorophenol, 5-chloro-3-trifluoromethyl phenol, phenol, 3-chloro-5-trifluoromethyl | |
3-chloro-5-(trifluoromethyl)phenol |
Specifications
570391-18-3 | |
1.485 | |
1 g | |
3-chloro-5-trifluoromethyl phenol, 3-chloro-5-hydroxybenzotrifluoride, 3-chloro-5-trifluoromethyl-phenol, 3-trifluoromethyl-5-chlorophenol, 5-chloro-3-trifluoromethyl phenol, phenol, 3-chloro-5-trifluoromethyl | |
OC1=CC(Cl)=CC(=C1)C(F)(F)F | |
196.55 | |
196.56 | |
3-Chloro-5-(trifluoromethyl)phenol |
C7H4ClF3O | |
MFCD04115852 | |
UN3265 | |
BPJWMOPEHPOKFT-UHFFFAOYSA-N | |
3-chloro-5-(trifluoromethyl)phenol | |
3852718 | |
97% |
Safety and Handling
GHS H Statement
H314-H318-H302-H312
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302+H312-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures