missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Chloro-5-(trifluoromethyl)pyridine-2-carboxylic acid, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H3425803

 View more versions of this product

Catalog No. AAH3425803


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-Chloro-5-(trifluoromethyl)pyridine-2-carboxylic acid
141°C to 143°C
MFCD00277482
3-chloro-5-trifluoromethyl pyridine-2-carboxylic acid, 3-chloro-5-trifluoromethyl picolinic acid, 2-pyridinecarboxylic acid, 3-chloro-5-trifluoromethyl, 3-chloro-5-trifluoromethyl-pyridine-2-carboxylic acid, 3-chloro-5-trifluoromethyl-2-pyridinecarboxylic acid, 2-carboxy-3-chloro-5-trifluoromethyl pyridine, ksc446c7n, 3-chloro-5-trifluoromethylpicolinic acid, 3-chloro-5-trifluoromethyl-picolinic acid
C1=C(C=NC(=C1Cl)C(=O)O)C(F)(F)F
225.551
225.6
80194-68-9
C7H3ClF3NO2
1 g
HXRMCZBDTDCCOP-UHFFFAOYSA-N
3-chloro-5-(trifluoromethyl)pyridine-2-carboxylic acid
2821908
97%

Chemical Identifiers

80194-68-9
225.551
HXRMCZBDTDCCOP-UHFFFAOYSA-N
2821908
C1=C(C=NC(=C1Cl)C(=O)O)C(F)(F)F
C7H3ClF3NO2
MFCD00277482
3-chloro-5-trifluoromethyl pyridine-2-carboxylic acid, 3-chloro-5-trifluoromethyl picolinic acid, 2-pyridinecarboxylic acid, 3-chloro-5-trifluoromethyl, 3-chloro-5-trifluoromethyl-pyridine-2-carboxylic acid, 3-chloro-5-trifluoromethyl-2-pyridinecarboxylic acid, 2-carboxy-3-chloro-5-trifluoromethyl pyridine, ksc446c7n, 3-chloro-5-trifluoromethylpicolinic acid, 3-chloro-5-trifluoromethyl-picolinic acid
3-chloro-5-(trifluoromethyl)pyridine-2-carboxylic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only