Learn More
3-Chloro-6-methylpyridazine 98.0+%, TCI America™
Supplier:  TCI America C227425G
 View more versions of this product
Catalog No. C227425G
Only null left
Explore available promotions
Description
Description
Specifications
Specifications
3-Chloro-6-methylpyridazine
60°C
C5H5ClN2
25 g
PRORLQAJNJMGAR-UHFFFAOYSA-N
3-chloro-6-methylpyridazine
227254
≥98.0% (GC,T)
1121-79-5
White-Yellow
MFCD00052905
3-chloro-6-methyl-pyridazine, 3-chloro-6-methyl pyridazine, pyridazine, 3-chloro-6-methyl, 6-chloro-3-methylpyridazine, pubchem10467, 3-methyl-6-chloropyridazine, 6-methyl-3-chloropyridazine, 3-chloro-6-methyl-1,2-diazine, 3-chloro-6-methylpyridazine
CC1=NN=C(C=C1)Cl
128.559
128.56
Crystalline Powder
Chemical Identifiers
1121-79-5
128.559
PRORLQAJNJMGAR-UHFFFAOYSA-N
227254
CC1=NN=C(C=C1)Cl
C5H5ClN2
MFCD00052905
3-chloro-6-methyl-pyridazine, 3-chloro-6-methyl pyridazine, pyridazine, 3-chloro-6-methyl, 6-chloro-3-methylpyridazine, pubchem10467, 3-methyl-6-chloropyridazine, 6-methyl-3-chloropyridazine, 3-chloro-6-methyl-1,2-diazine, 3-chloro-6-methylpyridazine
3-chloro-6-methylpyridazine
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
TSCA : No