missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Chloro-N-methylaniline, 97%, Thermo Scientific Chemicals

$70.33 - $222.85

Chemical Identifiers

CAS 7006-52-2
Molecular Formula C7H8ClN
Molecular Weight (g/mol) 141.60
MDL Number MFCD00052014
InChI Key WFGYSQDPURFIFL-UHFFFAOYSA-N
Synonym n-methyl-3-chloroaniline, 3-chloro-n-methylbenzenamine, benzenamine, 3-chloro-n-methyl, 3-chloro-phenyl-methyl-amine, n-methyl-m-chloroaniline, n1-methyl-3-chloroaniline, pubchem9202, acmc-209ocl, 3-chloro-1-methylaniline, 3-chloro-n-methyl aniline
PubChem CID 138900
IUPAC Name 3-chloro-N-methylaniline
SMILES CNC1=CC=CC(Cl)=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1241803
View Documents
Thermo Scientific Chemicals
A1241803
1 g
Each for $70.33
 
AAA1241806
View Documents
Thermo Scientific Chemicals
A1241806
5 g
Each for $222.85
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7006-52-2
141.60
WFGYSQDPURFIFL-UHFFFAOYSA-N
138900
CNC1=CC=CC(Cl)=C1
C7H8ClN
MFCD00052014
n-methyl-3-chloroaniline, 3-chloro-n-methylbenzenamine, benzenamine, 3-chloro-n-methyl, 3-chloro-phenyl-methyl-amine, n-methyl-m-chloroaniline, n1-methyl-3-chloroaniline, pubchem9202, acmc-209ocl, 3-chloro-1-methylaniline, 3-chloro-n-methyl aniline
3-chloro-N-methylaniline

Specifications

7006-52-2
243°C to 244°C
C7H8ClN
MFCD00052014
UN2810
n-methyl-3-chloroaniline, 3-chloro-n-methylbenzenamine, benzenamine, 3-chloro-n-methyl, 3-chloro-phenyl-methyl-amine, n-methyl-m-chloroaniline, n1-methyl-3-chloroaniline, pubchem9202, acmc-209ocl, 3-chloro-1-methylaniline, 3-chloro-n-methyl aniline
CNC1=CC=CC(Cl)=C1
141.60
141.6
3-Chloro-N-methylaniline
1.163
>110°C (230°F)
1.583
1 g
2802319
WFGYSQDPURFIFL-UHFFFAOYSA-N
3-chloro-N-methylaniline
138900
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H373-H302-H312-H315-H319
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only