Learn More
3-Chloro-N,N-dimethylaniline, 95%, Thermo Scientific Chemicals
$315.32 - $1128.38
Chemical Identifiers
CAS | 6848-13-1 |
---|---|
Molecular Formula | C8H10ClN |
Molecular Weight (g/mol) | 155.625 |
MDL Number | MFCD00018456 |
InChI Key | CHHCCYVOJBBCIY-UHFFFAOYSA-N |
Synonym | n,n-dimethyl-3-chloroaniline, 3-chloro-nn-dimethylaniline, benzenamine, 3-chloro-n,n-dimethyl, m-chloro-n,n-dimethylaniline, benzenamine,3-chloro-n,n-dimethyl, 3-chlorophenyl dimethylamine, pubchem3637, 3-chloranyl-n,n-dimethyl-aniline, 3-n,n-dimethylamino phenyl chloride |
PubChem CID | 23285 |
IUPAC Name | 3-chloro-N,N-dimethylaniline |
SMILES | CN(C)C1=CC(=CC=C1)Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL0856306
|
Thermo Scientific Chemicals
L0856306 |
5 g |
Each for $315.32
|
|
|||||
AAL0856314
|
Thermo Scientific Chemicals
L0856314 |
25 g |
Each for $1,128.38
|
|
|||||
Description
It can be used to produce 3-(3-dimethylamino-phenyl)-acrylic acid butyl ester at the temperature of 140°C in the presence of reagent Na2CO3 and solvent N,N-dimethyl-acetamide with the reaction time of 20 hours.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt can be used to produce 3-(3-dimethylamino-phenyl)-acrylic acid butyl ester at the temperature of 140°C in the presence of reagent Na2CO3 and solvent N,N-dimethyl-acetamide with the reaction time of 20 hours.
Solubility
Not miscible or difficult to mix in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
6848-13-1 | |
155.625 | |
CHHCCYVOJBBCIY-UHFFFAOYSA-N | |
23285 | |
CN(C)C1=CC(=CC=C1)Cl |
C8H10ClN | |
MFCD00018456 | |
n,n-dimethyl-3-chloroaniline, 3-chloro-nn-dimethylaniline, benzenamine, 3-chloro-n,n-dimethyl, m-chloro-n,n-dimethylaniline, benzenamine,3-chloro-n,n-dimethyl, 3-chlorophenyl dimethylamine, pubchem3637, 3-chloranyl-n,n-dimethyl-aniline, 3-n,n-dimethylamino phenyl chloride | |
3-chloro-N,N-dimethylaniline |
Specifications
6848-13-1 | |
C8H10ClN | |
MFCD00018456 | |
UN2810 | |
n,n-dimethyl-3-chloroaniline, 3-chloro-nn-dimethylaniline, benzenamine, 3-chloro-n,n-dimethyl, m-chloro-n,n-dimethylaniline, benzenamine,3-chloro-n,n-dimethyl, 3-chlorophenyl dimethylamine, pubchem3637, 3-chloranyl-n,n-dimethyl-aniline, 3-n,n-dimethylamino phenyl chloride | |
CHHCCYVOJBBCIY-UHFFFAOYSA-N | |
3-chloro-N,N-dimethylaniline | |
23285 | |
95% |
230°C to 232°C | |
1.576 | |
5 g | |
2206152 | |
Not miscible or difficult to mix in water. | |
CN(C)C1=CC(=CC=C1)Cl | |
155.625 | |
155.63 | |
3-Chloro-N,N-dimethylaniline |
Safety and Handling
GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber : 229-935-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only