missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Chloroaniline 99.0+%, TCI America™
$35.56 - $183.68
Chemical Identifiers
CAS | 108-42-9 |
---|---|
Molecular Formula | C6H6ClN |
Molecular Weight (g/mol) | 127.57 |
MDL Number | MFCD00007765 |
InChI Key | PNPCRKVUWYDDST-UHFFFAOYSA-N |
Synonym | m-chloroaniline, 3-chlorobenzenamine, benzenamine, 3-chloro, m-chloraniline, 3-chlorophenylamine, orange gc base, m-chlorophenylamine, m-aminochlorobenzene, m-chloroaminobenzene, aniline, m-chloro |
PubChem CID | 7932 |
IUPAC Name | 3-chloroaniline |
SMILES | NC1=CC=CC(Cl)=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
C011025ML
|
TCI America
C011025ML |
25 mL |
Each for $35.56
|
|
||||||
C0110500ML
|
TCI America
C0110500ML |
500 mL |
Each for $183.68
|
|
||||||
Chemical Identifiers
108-42-9 | |
127.57 | |
PNPCRKVUWYDDST-UHFFFAOYSA-N | |
7932 | |
NC1=CC=CC(Cl)=C1 |
C6H6ClN | |
MFCD00007765 | |
m-chloroaniline, 3-chlorobenzenamine, benzenamine, 3-chloro, m-chloraniline, 3-chlorophenylamine, orange gc base, m-chlorophenylamine, m-aminochlorobenzene, m-chloroaminobenzene, aniline, m-chloro | |
3-chloroaniline |
Specifications
-10°C | |
Yellow | |
25 mL | |
C6H6ClN | |
2019 | |
PNPCRKVUWYDDST-UHFFFAOYSA-N | |
3-chloroaniline | |
7932 | |
≥99.0% (GC) |
108-42-9 | |
230°C | |
Clear Liquid at 20°C | |
MFCD00007765 | |
m-chloroaniline, 3-chlorobenzenamine, benzenamine, 3-chloro, m-chloraniline, 3-chlorophenylamine, orange gc base, m-chlorophenylamine, m-aminochlorobenzene, m-chloroaminobenzene, aniline, m-chloro | |
NC1=CC=CC(Cl)=C1 | |
127.57 | |
127.57 | |
3-Chloroaniline |
Safety and Handling
EINECSNumber : (3)-0194
RTECSNumber : BX0350000
TSCA : Yes