Learn More
3-Chlorobenzenesulfonyl chloride, 98%, Thermo Scientific Chemicals
$61.49 - $604.34
Chemical Identifiers
CAS | 2888-06-4 |
---|---|
Molecular Formula | C6H4Cl2O2S |
Molecular Weight (g/mol) | 211.06 |
MDL Number | MFCD00051697 |
InChI Key | OINWZUJVEXUHCC-UHFFFAOYSA-N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1028303
|
Thermo Scientific Chemicals
A1028303 |
1 g |
Each for $61.49
|
|
|||||
AAA1028306
|
Thermo Scientific Chemicals
A1028306 |
5 g |
Each for $190.23
|
|
|||||
AAA1028314
|
Thermo Scientific Chemicals
A1028314 |
25 g |
Each for $604.34
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2888-06-4 | |
211.06 | |
OINWZUJVEXUHCC-UHFFFAOYSA-N |
C6H4Cl2O2S | |
MFCD00051697 |
Specifications
2888-06-4 | |
1.499 | |
>110°C (230°F) | |
1.569 | |
1 g | |
2937139 | |
3-chlorobenzene-1-sulfonyl chloride, 3-chlorobenzenesulphonyl chloride, 3-chlorobenzenesulfonylchloride, benzenesulfonyl chloride, 3-chloro, m-chlorobenzenesulfonyl chloride, 3-chloro-benzenesulfonyl chloride, 3-chlorophenylsulfonyl chloride, pubchem5076, m-chlorbenzolsulfochlorid, acmc-1cj4b | |
C1=CC(=CC(=C1)Cl)S(=O)(=O)Cl | |
211.06 | |
211.07 | |
3-Chlorobenzenesulfonyl chloride |
∼21°C | |
102°C to 104°C (1 mmHg) | |
C6H4Cl2O2S | |
MFCD00051697 | |
UN3265 | |
Moisture sensitive | |
OINWZUJVEXUHCC-UHFFFAOYSA-N | |
3-chlorobenzenesulfonyl chloride | |
17909 | |
98% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 220-753-0
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only