missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-chlorobenzenesulfonyl chloride, 98%, Thermo Scientific™

Chemical Identifiers

CAS 2888-06-4
Molecular Formula C6H4Cl2O2S
Molecular Weight (g/mol) 211.07
MDL Number MFCD00051697
InChI Key OINWZUJVEXUHCC-UHFFFAOYSA-N
Synonym 3-chlorobenzene-1-sulfonyl chloride, 3-chlorobenzenesulphonyl chloride, 3-chlorobenzenesulfonylchloride, benzenesulfonyl chloride, 3-chloro, m-chlorobenzenesulfonyl chloride, 3-chloro-benzenesulfonyl chloride, 3-chlorophenylsulfonyl chloride, pubchem5076, m-chlorbenzolsulfochlorid, acmc-1cj4b
PubChem CID 17909
IUPAC Name 3-chlorobenzenesulfonyl chloride
SMILES C1=CC(=CC(=C1)Cl)S(=O)(=O)Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC356480010
SDS View Documents Promotion Details
Thermo Scientific Chemicals
356480010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC356480050
SDS View Documents Promotion Details
Thermo Scientific Chemicals
356480050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC356480250
SDS View Documents Promotion Details
Thermo Scientific Chemicals
356480250
25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

2888-06-4
211.07
OINWZUJVEXUHCC-UHFFFAOYSA-N
17909
C1=CC(=CC(=C1)Cl)S(=O)(=O)Cl
C6H4Cl2O2S
MFCD00051697
3-chlorobenzene-1-sulfonyl chloride, 3-chlorobenzenesulphonyl chloride, 3-chlorobenzenesulfonylchloride, benzenesulfonyl chloride, 3-chloro, m-chlorobenzenesulfonyl chloride, 3-chloro-benzenesulfonyl chloride, 3-chlorophenylsulfonyl chloride, pubchem5076, m-chlorbenzolsulfochlorid, acmc-1cj4b
3-chlorobenzenesulfonyl chloride

Specifications

2888-06-4
100.0
102°C (0.9mmHg)
98%
C6H4Cl2O2S
ClC6H4SO2Cl
1g
3-chlorobenzene-1-sulfonyl chloride, 3-chlorobenzenesulphonyl chloride, 3-chlorobenzenesulfonylchloride, benzenesulfonyl chloride, 3-chloro, m-chlorobenzenesulfonyl chloride, 3-chloro-benzenesulfonyl chloride, 3-chlorophenylsulfonyl chloride, pubchem5076, m-chlorbenzolsulfochlorid, acmc-1cj4b
C1=CC(=CC(=C1)Cl)S(=O)(=O)Cl
211.07
211.07
3-chlorobenzenesulfonyl chloride
97.5
1.4990g/mL
Authentic
Glass bottle
1.5670 to 1.569
MFCD00051697
1.499
OINWZUJVEXUHCC-UHFFFAOYSA-N
3-chlorobenzenesulfonyl chloride
17909
98%
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 220-753-

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only