missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Chlorobenzo[b]thiophene-2-carbonyl chloride, 97%, Thermo Scientific™
Chemical Identifiers
CAS | 21815-91-8 |
---|---|
Molecular Formula | C9H4Cl2OS |
Molecular Weight (g/mol) | 231.09 |
MDL Number | MFCD00053069 |
InChI Key | GWKSSMDJEWPKCM-UHFFFAOYSA-N |
Synonym | 3-chlorobenzo b thiophene-2-carbonyl chloride, 3-chlorobenzothiophene-2-carbonyl chloride, 3-chloro-benzo b thiophene-2-carbonyl chloride, 3-chlorobenzo b thiophen-2-carbonyl chloride, benzo b thiophene-2-carbonyl chloride, 3-chloro, 3-chlorobenzo b-2-thiophenecarboxylic acid chloride, acmc-1cne6, 3-chlorobenzo b thiophene-2-carbonylchloride, 2-chlorocarbonyl-3-chlorobenzo b thiophene, 3-chlorobenzo thiophene-2-carbonyl chloride |
PubChem CID | 519898 |
IUPAC Name | 3-chloro-1-benzothiophene-2-carbonyl chloride |
SMILES | ClC(=O)C1=C(Cl)C2=CC=CC=C2S1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
BTB00300DA
|
Thermo Scientific Chemicals
BTB00300DA |
1g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
BTB00300EA
|
Thermo Scientific Chemicals
BTB00300EA |
10g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
BTB00300EB
|
Thermo Scientific Chemicals
BTB00300EB |
25g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
21815-91-8 | |
231.09 | |
GWKSSMDJEWPKCM-UHFFFAOYSA-N | |
519898 | |
ClC(=O)C1=C(Cl)C2=CC=CC=C2S1 |
C9H4Cl2OS | |
MFCD00053069 | |
3-chlorobenzo b thiophene-2-carbonyl chloride, 3-chlorobenzothiophene-2-carbonyl chloride, 3-chloro-benzo b thiophene-2-carbonyl chloride, 3-chlorobenzo b thiophen-2-carbonyl chloride, benzo b thiophene-2-carbonyl chloride, 3-chloro, 3-chlorobenzo b-2-thiophenecarboxylic acid chloride, acmc-1cne6, 3-chlorobenzo b thiophene-2-carbonylchloride, 2-chlorocarbonyl-3-chlorobenzo b thiophene, 3-chlorobenzo thiophene-2-carbonyl chloride | |
3-chloro-1-benzothiophene-2-carbonyl chloride |
Specifications
21815-91-8 | |
Amber Glass Bottle | |
MFCD00053069 | |
3-chlorobenzo b thiophene-2-carbonyl chloride, 3-chlorobenzothiophene-2-carbonyl chloride, 3-chloro-benzo b thiophene-2-carbonyl chloride, 3-chlorobenzo b thiophen-2-carbonyl chloride, benzo b thiophene-2-carbonyl chloride, 3-chloro, 3-chlorobenzo b-2-thiophenecarboxylic acid chloride, acmc-1cne6, 3-chlorobenzo b thiophene-2-carbonylchloride, 2-chlorocarbonyl-3-chlorobenzo b thiophene, 3-chlorobenzo thiophene-2-carbonyl chloride | |
ClC(=O)C1=C(Cl)C2=CC=CC=C2S1 | |
231.09 | |
231.1 |
97% | |
C9H4Cl2OS | |
1g | |
GWKSSMDJEWPKCM-UHFFFAOYSA-N | |
3-chloro-1-benzothiophene-2-carbonyl chloride | |
519898 | |
3-Chlorobenzo[b]thiophene-2-carbonyl chloride |