missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Chlorobenzophenone, 98+%, Thermo Scientific Chemicals

$66.48 - $445.59

Chemical Identifiers

CAS 1016-78-0
Molecular Formula C13H9ClO
Molecular Weight (g/mol) 216.664
MDL Number MFCD00009816
InChI Key CPLWKNRPZVNELG-UHFFFAOYSA-N
Synonym 3-chlorobenzophenone, 3-chlorophenyl phenyl methanone, benzophenone, 3-chloro, methanone, 3-chlorophenyl phenyl, m-chlorobenzophenone, 3-chlorophenyl-phenylmethanone, methanone, 3-chlorophenyl pheny, 3-chlorophenyl phenyl ketone, 5-chlorobenzophenone, pubchem24414
PubChem CID 66098
IUPAC Name (3-chlorophenyl)-phenylmethanone
SMILES C1=CC=C(C=C1)C(=O)C2=CC(=CC=C2)Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2308506
View Documents
Thermo Scientific Chemicals
B2308506
5 g
Each for $66.48
 
AAB2308514
View Documents
Thermo Scientific Chemicals
B2308514
25 g
Each for $155.05
 
AAB2308522
View Documents
Thermo Scientific Chemicals
B2308522
100 g
Each for $445.59
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1016-78-0
216.664
CPLWKNRPZVNELG-UHFFFAOYSA-N
66098
C1=CC=C(C=C1)C(=O)C2=CC(=CC=C2)Cl
C13H9ClO
MFCD00009816
3-chlorobenzophenone, 3-chlorophenyl phenyl methanone, benzophenone, 3-chloro, methanone, 3-chlorophenyl phenyl, m-chlorobenzophenone, 3-chlorophenyl-phenylmethanone, methanone, 3-chlorophenyl pheny, 3-chlorophenyl phenyl ketone, 5-chlorobenzophenone, pubchem24414
(3-chlorophenyl)-phenylmethanone

Specifications

1016-78-0
C13H9ClO
5 g
3-chlorobenzophenone, 3-chlorophenyl phenyl methanone, benzophenone, 3-chloro, methanone, 3-chlorophenyl phenyl, m-chlorobenzophenone, 3-chlorophenyl-phenylmethanone, methanone, 3-chlorophenyl pheny, 3-chlorophenyl phenyl ketone, 5-chlorobenzophenone, pubchem24414
C1=CC=C(C=C1)C(=O)C2=CC(=CC=C2)Cl
216.664
216.67
3-Chlorobenzophenone
81°C to 85°C
MFCD00009816
1869822
CPLWKNRPZVNELG-UHFFFAOYSA-N
(3-chlorophenyl)-phenylmethanone
66098
≥98%
Videos
Safety and Handling

Safety and Handling

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

EINECSNumber : 213-809-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only