Learn More
3-Chlorobenzyl bromide, 97%
CAS: 766-80-3 | C7H6BrCl | 205.48 g/mol
$58.62 - $192.42
Chemical Identifiers
| CAS | 766-80-3 |
|---|---|
| Molecular Formula | C7H6BrCl |
| Molecular Weight (g/mol) | 205.48 |
| MDL Number | MFCD00000597 |
| InChI Key | LZIYAIRGDHSVED-UHFFFAOYSA-N |
| Synonym | 3-chlorobenzyl bromide, 1-bromomethyl-3-chlorobenzene, 3-chlorobenzylbromide, alpha-bromo-3-chlorotoluene, m-chlorobenzyl bromide, benzene, 1-bromomethyl-3-chloro, alpha-bromo-m-chlorotoluene, .alpha.-bromo-m-chlorotoluene, 1-bromomethyl-3-chloro-benzene, 3-chorobenzylbromide |
| PubChem CID | 69838 |
| IUPAC Name | 1-(bromomethyl)-3-chlorobenzene |
| SMILES | ClC1=CC=CC(CBr)=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1584409
|
Thermo Scientific Chemicals
A1584409 |
10 g |
Each for $58.62
|
|
|||||
|
AAA1584418
|
Thermo Scientific Chemicals
A1584418 |
50 g |
Each for $192.42
|
|
|||||
Description
3-Chlorobenzyl bromide was used in the synthesis of symmetrical and unsymmetrical benzyl thioethers. It was used as starting reagent during the synthesis of 1-(3-chlorobenzyl)-2-(pyrrolidin-1-ylmethyl)-1H-benzimidazole dihydrochloride.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Chlorobenzyl bromide was used in the synthesis of symmetrical and unsymmetrical benzyl thioethers. It was used as starting reagent during the synthesis of 1-(3-chlorobenzyl)-2-(pyrrolidin-1-ylmethyl)-1H-benzimidazole dihydrochloride.
Solubility
Decomposes in water.
Notes
Store at room temperature. Keep away from oxidizing agents.
Chemical Identifiers
| 766-80-3 | |
| 205.48 | |
| LZIYAIRGDHSVED-UHFFFAOYSA-N | |
| 69838 | |
| ClC1=CC=CC(CBr)=C1 |
| C7H6BrCl | |
| MFCD00000597 | |
| 3-chlorobenzyl bromide, 1-bromomethyl-3-chlorobenzene, 3-chlorobenzylbromide, alpha-bromo-3-chlorotoluene, m-chlorobenzyl bromide, benzene, 1-bromomethyl-3-chloro, alpha-bromo-m-chlorotoluene, .alpha.-bromo-m-chlorotoluene, 1-bromomethyl-3-chloro-benzene, 3-chorobenzylbromide | |
| 1-(bromomethyl)-3-chlorobenzene |
Specifications
| 766-80-3 | |
| 108°C to 110°C (10 mmHg) | |
| Pungent | |
| 1.589 | |
| 10 g | |
| 636504 | |
| Decomposes in water. | |
| ClC1=CC=CC(CBr)=C1 | |
| 205.48 | |
| 205.49 | |
| 3-Chlorobenzyl bromide |
| 1.567 | |
| >110°C (230°F) | |
| C7H6BrCl | |
| MFCD00000597 | |
| UN3265 | |
| 3-chlorobenzyl bromide, 1-bromomethyl-3-chlorobenzene, 3-chlorobenzylbromide, alpha-bromo-3-chlorotoluene, m-chlorobenzyl bromide, benzene, 1-bromomethyl-3-chloro, alpha-bromo-m-chlorotoluene, .alpha.-bromo-m-chlorotoluene, 1-bromomethyl-3-chloro-benzene, 3-chorobenzylbromide | |
| LZIYAIRGDHSVED-UHFFFAOYSA-N | |
| 1-(bromomethyl)-3-chlorobenzene | |
| 69838 | |
| 97% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 212-171-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only