missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Chlorohexane 95.0+%, TCI America™

Supplier:  TCI America C090110ML

Catalog No. C090110ML

Explore available promotions

Description

Description

Specifications

Specifications

3-Chlorohexane
Yellow
C6H13Cl
10 mL
3-Hexyl Chloride
CCCC(CC)Cl
120.62
120.62
Liquid
2346-81-8
123°C
MFCD00039350
1993
BXSMMAVTEURRGG-UHFFFAOYSA-N
3-chlorohexane
33016
≥95.0% (GC)

Chemical Identifiers

2346-81-8
120.62
BXSMMAVTEURRGG-UHFFFAOYSA-N
33016
CCCC(CC)Cl
C6H13Cl
MFCD00039350
3-Hexyl Chloride
3-chlorohexane
Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions