missing translation for 'onlineSavingsMsg'
Learn More

3-(Chloromethyl)benzoyl Chloride 93.0+%, TCI America™

$76.43 - $231.14

Chemical Identifiers

CAS 63024-77-1
Molecular Formula C8H6Cl2O
Molecular Weight (g/mol) 189.035
MDL Number MFCD00000682
InChI Key YCAIYRWHKSJKEB-UHFFFAOYSA-N
Synonym 3-chloromethyl benzoyl chloride, 3-chloromethyl benzoylchloride, 3-chloromethyl-benzoyl chloride, benzoyl chloride, 3-chloromethyl, pubchem16448, 3-chlormethylbenzoylchloride, 3-chloromethylbenzoylchloride, acmc-1b65n
PubChem CID 2733324
IUPAC Name 3-(chloromethyl)benzoyl chloride
SMILES C1=CC(=CC(=C1)CCl)C(=O)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
C23145G
View Documents
TCI America
C23145G
5 g
Each for $76.43
Only null left
 
C231425G
missing translation for 'viewDocuments'
TCI America
C231425G
25 g
Each for $231.14
Only null left
 
Description

Description

Specifications

Chemical Identifiers

63024-77-1
189.035
YCAIYRWHKSJKEB-UHFFFAOYSA-N
2733324
C1=CC(=CC(=C1)CCl)C(=O)Cl
C8H6Cl2O
MFCD00000682
3-chloromethyl benzoyl chloride, 3-chloromethyl benzoylchloride, 3-chloromethyl-benzoyl chloride, benzoyl chloride, 3-chloromethyl, pubchem16448, 3-chlormethylbenzoylchloride, 3-chloromethylbenzoylchloride, acmc-1b65n
3-(chloromethyl)benzoyl chloride

Specifications

63024-77-1
Yellow
C8H6Cl2O
5 g
3-chloromethyl benzoyl chloride, 3-chloromethyl benzoylchloride, 3-chloromethyl-benzoyl chloride, benzoyl chloride, 3-chloromethyl, pubchem16448, 3-chlormethylbenzoylchloride, 3-chloromethylbenzoylchloride, acmc-1b65n
C1=CC(=CC(=C1)CCl)C(=O)Cl
189.035
189.04
Liquid
4°C
150°C
MFCD00000682
3265
YCAIYRWHKSJKEB-UHFFFAOYSA-N
3-(chloromethyl)benzoyl chloride
2733324
≥93.0% (GC)
3-(Chloromethyl)benzoyl Chloride
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents