missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Chloropropionyl chloride, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 109991000

 View more versions of this product

Catalog No. AC109991000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-Chloropropionyl chloride
625-36-5
100.0
Brown to Colorless
143.0°C to 145.0°C
Authentic
Glass bottle
1.4560 to 1.4580
MFCD00000747
02, 250
3-chloropropionyl chloride, propanoyl chloride, 3-chloro, propionyl chloride, 3-chloro, 3-chloropropionylchloride, beta-chloropropionyl chloride, 3-chloropropionic acid chloride, chloropropionyl chloride, propanoylchloride, 3-chloro, .beta.-chloropropanoyl chloride, .beta.-chloropropionyl chloride
INUNLMUAPJVRME-UHFFFAOYSA-N
3-chloropropanoyl chloride
69364
126.97
Liquid
98%
97.5
-32.0°C
1.3300g/mL
49°C
97.5% min. (GC)
C3H4Cl2O
ClCH2CH2COCl
100 mL
1.33
Solubility in water: reacts. Other solubilities: reacts violently with alcohol,amines,other,nucleophtilic solvents
ClCCC(Cl)=O
126.96
1.4 mPa.s (20°C)
98%

Chemical Identifiers

625-36-5
126.96
INUNLMUAPJVRME-UHFFFAOYSA-N
69364
ClCCC(Cl)=O
C3H4Cl2O
MFCD00000747
3-chloropropionyl chloride, propanoyl chloride, 3-chloro, propionyl chloride, 3-chloro, 3-chloropropionylchloride, beta-chloropropionyl chloride, 3-chloropropionic acid chloride, chloropropionyl chloride, propanoylchloride, 3-chloro, .beta.-chloropropanoyl chloride, .beta.-chloropropionyl chloride
3-chloropropanoyl chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Fatal if inhaled.
Harmful if swallowed.
Flammable liquid and vapor.
May be corrosive to metals.
Corrosive to the respiratory tract.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a POISON CENTER or doctor/physi

GHS Signal Word: Danger

EINECSNumber : 210-890-4

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only