missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Chloropropiophenone, 96%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1229009

 View more versions of this product

Catalog No. AAA1229009

Explore available promotions

Description

Description

3-Chloropropiophenone was used in the asymmetric reduction of (S)-3-chloro-1-phenylpropanol using preheated Candida utilis cells immobilized in calcium alginate gel beads. It was also used in the synthesis of (R)-3-chloro-1-phenyl-1-propanol via asymmetric reduction using in-situ generated oxazaborolidine catalyst derived from (S)-α,α-diphenylprolinol. It is a precursor of phenyl vinyl ketone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Chloropropiophenone was used in the asymmetric reduction of (S)-3-chloro-1-phenylpropanol using preheated Candida utilis cells immobilized in calcium alginate gel beads. It was also used in the synthesis of (R)-3-chloro-1-phenyl-1-propanol via asymmetric reduction using in-situ generated oxazaborolidine catalyst derived from (S)-α,α-diphenylprolinol. It is a precursor of phenyl vinyl ketone.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Specifications

Specifications

3-Chloropropiophenone
46°C to 50°C
>110°C (230°F)
MFCD00000990
2043580
Insoluble in water.
C1=CC=C(C=C1)C(=O)CCCl
168.62
168.62
936-59-4
113°C to 115°C (4 mmHg)
C9H9ClO
10 g
3-chloropropiophenone, 1-propanone, 3-chloro-1-phenyl, beta-chloropropiophenone, 2-chloroethyl phenyl ketone, 3-chloropropanophenone, 3-chloro-1-phenyl-1-propanone, propiophenone, 3-chloro, 3-chloro-1-phenyl-propan-1-one, .beta.-chloropropiophenone, .omega.-chloropropiophenone
KTJRGPZVSKWRTJ-UHFFFAOYSA-N
3-chloro-1-phenylpropan-1-one
70295
96%

Chemical Identifiers

936-59-4
168.62
KTJRGPZVSKWRTJ-UHFFFAOYSA-N
70295
C1=CC=C(C=C1)C(=O)CCCl
C9H9ClO
MFCD00000990
3-chloropropiophenone, 1-propanone, 3-chloro-1-phenyl, beta-chloropropiophenone, 2-chloroethyl phenyl ketone, 3-chloropropanophenone, 3-chloro-1-phenyl-1-propanone, propiophenone, 3-chloro, 3-chloro-1-phenyl-propan-1-one, .beta.-chloropropiophenone, .omega.-chloropropiophenone
3-chloro-1-phenylpropan-1-one
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 213-317-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only