missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(3-Chloropropyl)benzene, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2044522

 View more versions of this product

Catalog No. AAB2044522


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

(3-Chloropropyl)benzene
1.05
87°C (189°F)
C6H5(CH2)3Cl
MFCD00001001
3-chloropropyl benzene, 1-chloro-3-phenylpropane, 3-phenylpropyl chloride, benzene, 3-chloropropyl, 3-phenyl-1-chloropropane, 3-chloro-1-phenylpropane, .gamma.-chloropropylbenzene, benzene, chloropropyl, .gamma.-phenylpropyl chloride, 3-penylpropylchloride
C1=CC=C(C=C1)CCCCl
154.637
154.64
104-52-9
218°C to 220°C
C9H11Cl
1.522
100 g
XZBXAYCCBFTQHH-UHFFFAOYSA-N
3-chloropropylbenzene
7706
98%

Chemical Identifiers

104-52-9
154.637
XZBXAYCCBFTQHH-UHFFFAOYSA-N
7706
C1=CC=C(C=C1)CCCCl
C9H11Cl
MFCD00001001
3-chloropropyl benzene, 1-chloro-3-phenylpropane, 3-phenylpropyl chloride, benzene, 3-chloropropyl, 3-phenyl-1-chloropropane, 3-chloro-1-phenylpropane, .gamma.-chloropropylbenzene, benzene, chloropropyl, .gamma.-phenylpropyl chloride, 3-penylpropylchloride
3-chloropropylbenzene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

EINECSNumber : 203-210-2

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only