missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(3-chloropropyl)trimethoxysilane, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 137280250

 View more versions of this product

Catalog No. AC137280250


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

(3-Chloropropyl)trimethoxysilane
98.0
Colorless to Yellow
195°C to 196°C (750.0 mmHg)
Authentic
Glass bottle
1.4190 to 1.421
MFCD00000997
1.07
Solubility in water: soluble
CO[Si](CCCCl)(OC)OC
198.72
198.72
Liquid
2530-87-2
100.0
1.0700g/mL
78°C
98% min. (GC)
C6H15ClO3Si
(CH3O)3Si(CH2)3Cl
25 mL
3-chloropropyl trimethoxysilane, silane, 3-chloropropyl trimethoxy, 3-chloropropyltrimethyoxysilane, sila-ace s 620, cps-m, silane 3-chloropropyl tris methoxy, 3-trimethoxysilyl propyl chloride, unii-t21bnl1s7f, cptmo, 3-chloropropyl trimethoxysilan
OXYZDRAJMHGSMW-UHFFFAOYSA-N
3-chloropropyl(trimethoxy)silane
62449
98+%

Chemical Identifiers

2530-87-2
198.72
OXYZDRAJMHGSMW-UHFFFAOYSA-N
62449
CO[Si](CCCCl)(OC)OC
C6H15ClO3Si
MFCD00000997
3-chloropropyl trimethoxysilane, silane, 3-chloropropyl trimethoxy, 3-chloropropyltrimethyoxysilane, sila-ace s 620, cps-m, silane 3-chloropropyl tris methoxy, 3-trimethoxysilyl propyl chloride, unii-t21bnl1s7f, cptmo, 3-chloropropyl trimethoxysilan
3-chloropropyl(trimethoxy)silane
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 219-787-9

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only