missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Chloropropyldichloromethylsilane 97.0+%, TCI America™
Supplier: TCI America D19955ML
Specifications
3-Chloropropyldichloromethylsilane | |
185°C | |
MFCD00000996 | |
2986 | |
UCJHMXXKIKBHQP-UHFFFAOYSA-N | |
dichloro-(3-chloropropyl)-methylsilane | |
82235 | |
≥97.0% (GC) |
7787-93-1 | |
C4H9Cl3Si | |
5 mL | |
3-(Dichloromethylsilyl)propyl Chloride | |
C[Si](CCCCl)(Cl)Cl | |
191.551 | |
191.55 | |
Liquid |
Chemical Identifiers
7787-93-1 | |
191.551 | |
UCJHMXXKIKBHQP-UHFFFAOYSA-N | |
82235 | |
C[Si](CCCCl)(Cl)Cl |
C4H9Cl3Si | |
MFCD00000996 | |
3-(Dichloromethylsilyl)propyl Chloride | |
dichloro-(3-chloropropyl)-methylsilane |
Safety and Handling
EINECSNumber : (2)-2047
TSCA : Yes