Learn More
(3-Cyanopropyl)triethoxysilane, 97%, Thermo Scientific Chemicals
$86.67 - $252.32
Chemical Identifiers
CAS | 1067-47-6 |
---|---|
Molecular Formula | C10H21NO3Si |
Molecular Weight (g/mol) | 231.367 |
MDL Number | MFCD00042636 |
InChI Key | VGIURMCNTDVGJM-UHFFFAOYSA-N |
Synonym | 3-cyanopropyltriethoxysilane, butanenitrile, 4-triethoxysilyl, 4-triethoxysilyl butanenitrile, 4-triethoxysilyl butyronitrile, butyronitrile, 4-triethoxysilyl, silane, triethoxy 3-cyanopropyl, triethoxy-3-kyanpropylsilan, unii-n78w6a24dx, 3-cyanopropyl triethoxysilane, triethoxy-3-kyanpropylsilan czech |
PubChem CID | 14024 |
IUPAC Name | 4-triethoxysilylbutanenitrile |
SMILES | CCO[Si](CCCC#N)(OCC)OCC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL0536714
|
Thermo Scientific Chemicals
L0536714 |
25 g |
Each for $86.67
|
|
|||||
AAL0536722
|
Thermo Scientific Chemicals
L0536722 |
100 g |
Each for $252.32
|
|
|||||
Description
(3-Cyanopropyl)triethoxysilane is used as cyanoalkyl silane coupling agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications(3-Cyanopropyl)triethoxysilane is used as cyanoalkyl silane coupling agent.
Solubility
Not miscible in water.
Notes
Moisture Sensitive. Incompatible materials are Oxidizing agents and water.
Chemical Identifiers
1067-47-6 | |
231.367 | |
VGIURMCNTDVGJM-UHFFFAOYSA-N | |
14024 | |
CCO[Si](CCCC#N)(OCC)OCC |
C10H21NO3Si | |
MFCD00042636 | |
3-cyanopropyltriethoxysilane, butanenitrile, 4-triethoxysilyl, 4-triethoxysilyl butanenitrile, 4-triethoxysilyl butyronitrile, butyronitrile, 4-triethoxysilyl, silane, triethoxy 3-cyanopropyl, triethoxy-3-kyanpropylsilan, unii-n78w6a24dx, 3-cyanopropyl triethoxysilane, triethoxy-3-kyanpropylsilan czech | |
4-triethoxysilylbutanenitrile |
Specifications
238°C to 241°C | |
25 g | |
NCCH2CH2CH2Si(OCH2CH3)3 | |
1942237 | |
3-cyanopropyltriethoxysilane, butanenitrile, 4-triethoxysilyl, 4-triethoxysilyl butanenitrile, 4-triethoxysilyl butyronitrile, butyronitrile, 4-triethoxysilyl, silane, triethoxy 3-cyanopropyl, triethoxy-3-kyanpropylsilan, unii-n78w6a24dx, 3-cyanopropyl triethoxysilane, triethoxy-3-kyanpropylsilan czech | |
VGIURMCNTDVGJM-UHFFFAOYSA-N | |
4-triethoxysilylbutanenitrile | |
14024 | |
97% | |
Moisture sensitive | |
(3-Cyanopropyl)triethoxysilane |
1067-47-6 | |
C10H21NO3Si | |
MFCD00042636 | |
74°C (165°F) | |
Not miscible in water. | |
CCO[Si](CCCC#N)(OCC)OCC | |
231.367 | |
231.37 | |
1.417 | |
0.961 |
Safety and Handling
GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
P210-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q-P501c
H227-H315-H319
EINECSNumber : 213-931-4
RTECSNumber : VV2785000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only