Learn More
3-Cyclohexyl-1-propyne, 97%, Thermo Scientific Chemicals
$80.14 - $254.18
Chemical Identifiers
CAS | 17715-00-3 |
---|---|
Molecular Formula | C9H14 |
Molecular Weight (g/mol) | 122.211 |
MDL Number | MFCD00041562 |
InChI Key | UARFKZSJGDQRLF-UHFFFAOYSA-N |
Synonym | 3-cyclohexyl-1-propyne, prop-2-yn-1-ylcyclohexane, 1-prop-2-ynylcyclohexane, 3-cyclohexylpropyne, 2-propynylcyclohexane, propyne, 3-cyclohexyl, cyclohexane, 2-propynyl-9ci, prop-2-ynyl-cyclohexane, cyclohexane, 2-propynyl, 3-cyclohexylprop-1-yne |
PubChem CID | 87265 |
IUPAC Name | prop-2-ynylcyclohexane |
SMILES | C#CCC1CCCCC1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2050503
|
Thermo Scientific Chemicals
B2050503 |
1 g |
Each for $80.14
|
|
|||||
AAB2050506
|
Thermo Scientific Chemicals
B2050506 |
5 g |
Each for $254.18
|
|
|||||
Description
3-Cyclohexyl-1-propyne, is used as a pharmaceutical intermediate. It can be used to produce (5-bromo-4,4,5,5-tetrafluoro-pent-2-enyl)-cyclohexane at the temperature of 40°C. It will need reagents (NH4)2S2O8, HCO2Na·2H2O and solvent dimethylformamide with the reaction time of 3 hours. It is also used as organic synthesis intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Cyclohexyl-1-propyne, is used as a pharmaceutical intermediate. It can be used to produce (5-bromo-4,4,5,5-tetrafluoro-pent-2-enyl)-cyclohexane at the temperature of 40°C. It will need reagents (NH4)2S2O8, HCO2Na·2H2O and solvent dimethylformamide with the reaction time of 3 hours. It is also used as organic synthesis intermediates.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Chemical Identifiers
17715-00-3 | |
122.211 | |
UARFKZSJGDQRLF-UHFFFAOYSA-N | |
87265 | |
C#CCC1CCCCC1 |
C9H14 | |
MFCD00041562 | |
3-cyclohexyl-1-propyne, prop-2-yn-1-ylcyclohexane, 1-prop-2-ynylcyclohexane, 3-cyclohexylpropyne, 2-propynylcyclohexane, propyne, 3-cyclohexyl, cyclohexane, 2-propynyl-9ci, prop-2-ynyl-cyclohexane, cyclohexane, 2-propynyl, 3-cyclohexylprop-1-yne | |
prop-2-ynylcyclohexane |
Specifications
17715-00-3 | |
157°C to 158°C | |
C9H14 | |
MFCD00041562 | |
UN3295 | |
3-cyclohexyl-1-propyne, prop-2-yn-1-ylcyclohexane, 1-prop-2-ynylcyclohexane, 3-cyclohexylpropyne, 2-propynylcyclohexane, propyne, 3-cyclohexyl, cyclohexane, 2-propynyl-9ci, prop-2-ynyl-cyclohexane, cyclohexane, 2-propynyl, 3-cyclohexylprop-1-yne | |
UARFKZSJGDQRLF-UHFFFAOYSA-N | |
prop-2-ynylcyclohexane | |
87265 | |
97% |
0.845 | |
35°C (95°F) | |
1.462 | |
1 g | |
1849402 | |
Slightly soluble in water. | |
C#CCC1CCCCC1 | |
122.211 | |
122.21 | |
3-Cyclohexyl-1-propyne |
Safety and Handling
GHS H Statement
H226
Flammable liquid and vapour.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H226
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
RTECSNumber : UK4725000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only