missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Cyclopentyl-L-alanine, 95%, Thermo Scientific Chemicals

$188.10 - $664.73

Chemical Identifiers

CAS 99295-82-6
Molecular Formula C8H15NO2
Molecular Weight (g/mol) 157.213
MDL Number MFCD01632069
InChI Key KDYAKYRBGLKMAK-ZETCQYMHSA-N
Synonym s-2-amino-3-cyclopentylpropanoic acid, h-cpa-oh, 3-cyclopentylalanine, l-3-cyclopentylalanine, 3-cyclopentane-l-alanine, 2s-2-amino-3-cyclopentylpropanoic acid, s-2-amino-3-cyclopentyl-propionic acid, s-alfa-amino-cyclopentanepropanoic acid ;3-cyclopentane-l-alanine, ambotzhaa1075, l-beta-cyclopentylalanine
PubChem CID 7000137
IUPAC Name (2S)-2-amino-3-cyclopentylpropanoic acid
SMILES C1CCC(C1)CC(C(=O)O)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH66600MD
View Documents
Thermo Scientific Chemicals
H66600MD
250 mg
Each for $188.10
 
AAH6660003
View Documents
Thermo Scientific Chemicals
H6660003
1 g
Each for $664.73
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

99295-82-6
157.213
KDYAKYRBGLKMAK-ZETCQYMHSA-N
7000137
C1CCC(C1)CC(C(=O)O)N
C8H15NO2
MFCD01632069
s-2-amino-3-cyclopentylpropanoic acid, h-cpa-oh, 3-cyclopentylalanine, l-3-cyclopentylalanine, 3-cyclopentane-l-alanine, 2s-2-amino-3-cyclopentylpropanoic acid, s-2-amino-3-cyclopentyl-propionic acid, s-alfa-amino-cyclopentanepropanoic acid ;3-cyclopentane-l-alanine, ambotzhaa1075, l-beta-cyclopentylalanine
(2S)-2-amino-3-cyclopentylpropanoic acid

Specifications

99295-82-6
C8H15NO2
10659781
KDYAKYRBGLKMAK-ZETCQYMHSA-N
(2S)-2-amino-3-cyclopentylpropanoic acid
157.213
157.21
Powder
White
MFCD01632069
s-2-amino-3-cyclopentylpropanoic acid, h-cpa-oh, 3-cyclopentylalanine, l-3-cyclopentylalanine, 3-cyclopentane-l-alanine, 2s-2-amino-3-cyclopentylpropanoic acid, s-2-amino-3-cyclopentyl-propionic acid, s-alfa-amino-cyclopentanepropanoic acid ;3-cyclopentane-l-alanine, ambotzhaa1075, l-beta-cyclopentylalanine
C1CCC(C1)CC(C(=O)O)N
250 mg
7000137
95%
3-Cyclopentyl-L-alanine
Videos
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C

SDS
Documents

Documents

RUO – Research Use Only