missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Cyclopentyloxy-4-methoxybenzyl alcohol, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H6244106
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
3-Cyclopentyloxy-4-methoxybenzyl alcohol | |
C13H18O3 | |
5 g | |
JIRHAGAOHOYLNO-UHFFFAOYSA-N | |
(3-cyclopentyloxy-4-methoxyphenyl)methanol | |
11138706 | |
97% |
133332-49-7 | |
MFCD03844627 | |
3-cyclopentyloxy-4-methoxybenzyl alcohol, 3-cyclopentyloxy-4-methoxyphenyl methanol, 3-cyclopentyloxy-4-methoxybenzylalcohol, acmc-1cioj, 3-cyclopentoxy-4-methoxybenzyl alcohol, 3-cyclopentyloxy-4-methoxy-benzyl alcohol | |
COC1=C(C=C(C=C1)CO)OC2CCCC2 | |
222.284 | |
222.28 |
Chemical Identifiers
133332-49-7 | |
222.284 | |
JIRHAGAOHOYLNO-UHFFFAOYSA-N | |
11138706 | |
COC1=C(C=C(C=C1)CO)OC2CCCC2 |
C13H18O3 | |
MFCD03844627 | |
3-cyclopentyloxy-4-methoxybenzyl alcohol, 3-cyclopentyloxy-4-methoxyphenyl methanol, 3-cyclopentyloxy-4-methoxybenzylalcohol, acmc-1cioj, 3-cyclopentoxy-4-methoxybenzyl alcohol, 3-cyclopentyloxy-4-methoxy-benzyl alcohol | |
(3-cyclopentyloxy-4-methoxyphenyl)methanol |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures