Learn More
3'-Deoxyadenosine, 98%, Thermo Scientific Chemicals
$328.25 - $893.15
Chemical Identifiers
CAS | 73-03-0 |
---|---|
Molecular Formula | C10H13N5O3 |
Molecular Weight (g/mol) | 251.25 |
MDL Number | MFCD00037998 |
InChI Key | OFEZSBMBBKLLBJ-PNFQIUPPNA-N |
Synonym | cordycepin, 3'-deoxyadenosine, cordycepine, 9-cordyceposidoadenine, adenosine, 3'-deoxy, 9-beta-d-3'-deoxyribofuranosyl adenine, unii-gz8vf4m2j8, 9h-purine, 6-amino-9-3-deoxy-beta-d-ribofuranosyl, ccris 3233, gz8vf4m2j8 |
PubChem CID | 6303 |
ChEBI | CHEBI:29014 |
SMILES | NC1=C2N=CN([C@@H]3O[C@H](CO)C[C@H]3O)C2=NC=N1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ65366MB
|
Thermo Scientific Chemicals
J65366MB |
25 mg |
Each for $328.25
|
|
|||||
AAJ65366MC
|
Thermo Scientific Chemicals
J65366MC |
100 mg |
Each for $893.15
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
73-03-0 | |
251.25 | |
OFEZSBMBBKLLBJ-PNFQIUPPNA-N | |
6303 | |
NC1=C2N=CN([C@@H]3O[C@H](CO)C[C@H]3O)C2=NC=N1 |
C10H13N5O3 | |
MFCD00037998 | |
cordycepin, 3'-deoxyadenosine, cordycepine, 9-cordyceposidoadenine, adenosine, 3'-deoxy, 9-beta-d-3'-deoxyribofuranosyl adenine, unii-gz8vf4m2j8, 9h-purine, 6-amino-9-3-deoxy-beta-d-ribofuranosyl, ccris 3233, gz8vf4m2j8 | |
CHEBI:29014 |
Specifications
73-03-0 | |
C10H13N5O3 | |
25 mg | |
35194 | |
cordycepin, 3'-deoxyadenosine, cordycepine, 9-cordyceposidoadenine, adenosine, 3'-deoxy, 9-beta-d-3'-deoxyribofuranosyl adenine, unii-gz8vf4m2j8, 9h-purine, 6-amino-9-3-deoxy-beta-d-ribofuranosyl, ccris 3233, gz8vf4m2j8 | |
NC1=C2N=CN([C@@H]3O[C@H](CO)C[C@H]3O)C2=NC=N1 | |
251.25 | |
CHEBI:29014 | |
98% | |
3'-Deoxyadenosine |
White | |
MFCD00037998 | |
UN2811 | |
14,2529 | |
OFEZSBMBBKLLBJ-PNFQIUPPNA-N | |
(2R,3R,5S)-2-(6-aminopurin-9-yl)-5-(hydroxymethyl)oxolan-3-ol | |
6303 | |
251.24 | |
Powder |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H301-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 200-791-4
RTECSNumber : AU7358610
TSCA : No
Recommended Storage : Store at -20°C
RUO – Research Use Only