Learn More
3-Diethylaminophenol, 99%
CAS: 91-68-9 | C10H15NO | 165.24 g/mol
Supplier: Thermo Scientific Chemicals 146631000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 3-Diethylaminophenol | |
| 91-68-9 | |
| 100.0 | |
| Brown-Gray or Red | |
| 141°C | |
| 98.5% min. (GC) | |
| C10H15NO | |
| MFCD00002265 | |
| 13, 408 | |
| Solubility in water: insoluble. Other solubilities: soluble in alcohols,ethers,ketones,esters,,carbon disulfide,caustic soda,alkalis and,aromatic hydrocarbons,insoluble in ligroin | |
| CCN(CC)C1=CC=CC(O)=C1 | |
| 165.24 | |
| 165.23 | |
| Flakes or Granules |
| 99% | |
| 98.5 | |
| 70°C to 73°C | |
| 170°C (15.0 mmHg) | |
| Authentic | |
| Glass bottle | |
| (C2H5)2NC6H4OH | |
| 100 g | |
| 3-diethylamino phenol, phenol, 3-diethylamino, n,n-diethyl-m-aminophenol, n,n-diethyl-3-aminophenol, m-diethylamino phenol, 3-hydroxy-n,n-diethylaniline, phenol, m-diethylamino, n,n-diethyl-3-hydroxyaniline, unii-bqy98q7yu5, ccris 4615 | |
| WAVOOWVINKGEHS-UHFFFAOYSA-N | |
| 3-(diethylamino)phenol | |
| 7062 | |
| 99% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Wash face,hands and any exposed skin thoroughly after handling.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not eat,drink or smoke wh
GHS Signal Word: Warning
EINECSNumber : 202-090-9
RTECSNumber : SL0550500
TSCA : TSCA
RUO – Research Use Only