missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-(Dimethylamino)propyl Chloride Hydrochloride 98.0+%, TCI America™
$37.52 - $156.03
Chemical Identifiers
CAS | 5407-04-5 |
---|---|
Molecular Formula | C5H13Cl2N |
Molecular Weight (g/mol) | 158.07 |
MDL Number | MFCD00012521 |
InChI Key | LJQNMDZRCXJETK-UHFFFAOYSA-N |
Synonym | 3-dimethylaminopropylchloride hydrochloride, 3-chloro-n,n-dimethylpropan-1-amine hydrochloride, 3-dimethylaminopropyl chloride hydrochloride, 3-dimethylamino propyl chloride hydrochloride, n,n-dimethyl-3-chloropropylamine hydrochloride, 3-chloropropyl dimethylamine hydrochloride, 3-chloro-n,n-dimethylpropylamine hydrochloride, dimethylaminopropyl chloride hydrochloride, ccris 7054, dimethylaminopropyl chloride, hydrochloride |
PubChem CID | 94308 |
IUPAC Name | 3-chloro-N,N-dimethylpropan-1-amine;hydrochloride |
SMILES | CN(C)CCCCl.Cl |
Chemical Identifiers
5407-04-5 | |
158.07 | |
LJQNMDZRCXJETK-UHFFFAOYSA-N | |
94308 | |
CN(C)CCCCl.Cl |
C5H13Cl2N | |
MFCD00012521 | |
3-dimethylaminopropylchloride hydrochloride, 3-chloro-n,n-dimethylpropan-1-amine hydrochloride, 3-dimethylaminopropyl chloride hydrochloride, 3-dimethylamino propyl chloride hydrochloride, n,n-dimethyl-3-chloropropylamine hydrochloride, 3-chloropropyl dimethylamine hydrochloride, 3-chloro-n,n-dimethylpropylamine hydrochloride, dimethylaminopropyl chloride hydrochloride, ccris 7054, dimethylaminopropyl chloride, hydrochloride | |
3-chloro-N,N-dimethylpropan-1-amine;hydrochloride |
Specifications
5407-04-5 | |
White | |
MFCD00012521 | |
3-dimethylaminopropylchloride hydrochloride, 3-chloro-n,n-dimethylpropan-1-amine hydrochloride, 3-dimethylaminopropyl chloride hydrochloride, 3-dimethylamino propyl chloride hydrochloride, n,n-dimethyl-3-chloropropylamine hydrochloride, 3-chloropropyl dimethylamine hydrochloride, 3-chloro-n,n-dimethylpropylamine hydrochloride, dimethylaminopropyl chloride hydrochloride, ccris 7054, dimethylaminopropyl chloride, hydrochloride | |
CN(C)CCCCl.Cl | |
158.07 | |
158.07 | |
Crystalline Powder |
143°C | |
C5H13Cl2N | |
25 g | |
LJQNMDZRCXJETK-UHFFFAOYSA-N | |
3-chloro-N,N-dimethylpropan-1-amine;hydrochloride | |
94308 | |
≥98.0% (T) | |
3-(Dimethylamino)propyl Chloride Hydrochloride |
Safety and Handling
EINECSNumber : (2)-0177&(9)-2009
RTECSNumber : UI0209950
TSCA : Yes