missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Dimethylaminophenol, 97+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2306706

 View more versions of this product

Catalog No. AAB2306706

Explore available promotions

Description

Description

3-Dimethylaminophenol can be used as neostigmine bromide intermediate and dye intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Dimethylaminophenol can be used as neostigmine bromide intermediate and dye intermediate.

Solubility
Slightly soluble in water.

Notes
Light sensitive. Store away from oxidizing agents, light.
Specifications

Specifications

3-Dimethylaminophenol
80°C to 84°C
148°C (298°F)
C8H11NO
5 g
Light sensitive
Slightly soluble in water.
CN(C)C1=CC=CC(O)=C1
137.18
137.18
99-07-0
267°C
Pungent
MFCD00002264
774631
3-dimethylamino phenol, phenol, 3-dimethylamino, m-dimethylamino phenol, m-dimethylaminophenol, n,n-dimethyl-m-aminophenol, n,n-dimethyl-3-aminophenol, phenol, m-dimethylamino, 3-hydroxyphenyl dimethylamine, 3-hydroxy-n,n-dimethylaniline, 3-n,n-dimethylaminophenol
MESJRHHDBDCQTH-UHFFFAOYSA-N
3-(dimethylamino)phenol
7421
≥97%

Chemical Identifiers

99-07-0
137.18
MESJRHHDBDCQTH-UHFFFAOYSA-N
7421
CN(C)C1=CC=CC(O)=C1
C8H11NO
MFCD00002264
3-dimethylamino phenol, phenol, 3-dimethylamino, m-dimethylamino phenol, m-dimethylaminophenol, n,n-dimethyl-m-aminophenol, n,n-dimethyl-3-aminophenol, phenol, m-dimethylamino, 3-hydroxyphenyl dimethylamine, 3-hydroxy-n,n-dimethylaniline, 3-n,n-dimethylaminophenol
3-(dimethylamino)phenol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

EINECSNumber : 202-727-0

RTECSNumber : SL1050000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only