Learn More
3-(Diphenylphosphino)propylamine, 97+%, Thermo Scientific Chemicals
$88.51 - $243.56
Chemical Identifiers
CAS | 16605-03-1 |
---|---|
Molecular Formula | C15H18NP |
Molecular Weight (g/mol) | 243.29 |
MDL Number | MFCD00233832 |
InChI Key | DQZWMOWSTWWMPP-UHFFFAOYSA-N |
Synonym | 3-diphenylphosphino-1-propylamine, 3-diphenylphosphino propan-1-amine, 3-aminopropyl diphenylphosphine, 3-diphenylphosphino propylamine, 3-diphenylphosphanyl-propylamine, 1-propanamine,3-diphenylphosphino, 3-aminopropyl diphenylphosphane, 3-diphenylphosphanyl propan-1-amine, acmc-20aniq, 3-aminopropyldiphenylphosphine |
PubChem CID | 280677 |
IUPAC Name | 3-diphenylphosphanylpropan-1-amine |
SMILES | NCCCP(C1=CC=CC=C1)C1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH60672MD
|
Thermo Scientific Chemicals
H60672MD |
250 mg |
Each for $88.51
|
|
|||||
AAH6067203
|
Thermo Scientific Chemicals
H6067203 |
1 g |
Each for $243.56
|
|
|||||
Description
3-(Diphenylphosphino)propylamine is used as a paharmaceutical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-(Diphenylphosphino)propylamine is used as a paharmaceutical.
Solubility
Not miscible or difficult to mix in water.
Notes
Air sensitive. Store under inert gas. Store away from oxidizing agents, air, acids.
Chemical Identifiers
16605-03-1 | |
243.29 | |
DQZWMOWSTWWMPP-UHFFFAOYSA-N | |
280677 | |
NCCCP(C1=CC=CC=C1)C1=CC=CC=C1 |
C15H18NP | |
MFCD00233832 | |
3-diphenylphosphino-1-propylamine, 3-diphenylphosphino propan-1-amine, 3-aminopropyl diphenylphosphine, 3-diphenylphosphino propylamine, 3-diphenylphosphanyl-propylamine, 1-propanamine,3-diphenylphosphino, 3-aminopropyl diphenylphosphane, 3-diphenylphosphanyl propan-1-amine, acmc-20aniq, 3-aminopropyldiphenylphosphine | |
3-diphenylphosphanylpropan-1-amine |
Specifications
16605-03-1 | |
(C6H5)2PCH2CH2CH2NH2 | |
250 mg | |
2730850 | |
3-diphenylphosphino-1-propylamine, 3-diphenylphosphino propan-1-amine, 3-aminopropyl diphenylphosphine, 3-diphenylphosphino propylamine, 3-diphenylphosphanyl-propylamine, 1-propanamine,3-diphenylphosphino, 3-aminopropyl diphenylphosphane, 3-diphenylphosphanyl propan-1-amine, acmc-20aniq, 3-aminopropyldiphenylphosphine | |
DQZWMOWSTWWMPP-UHFFFAOYSA-N | |
3-diphenylphosphanylpropan-1-amine | |
280677 | |
≥97% |
C15H18NP | |
MFCD00233832 | |
UN2735 | |
Air Sensitive | |
Not miscible or difficult to mix in water. | |
NCCCP(C1=CC=CC=C1)C1=CC=CC=C1 | |
243.29 | |
243.28 | |
3-(Diphenylphosphino)propylamine |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only