Learn More
3-Ethoxy-2-cyclohexen-1-one, 99%
CAS: 5323-87-5 | C8H12O2 | 140.18 g/mol
$87.90 - $1126.26
Chemical Identifiers
| CAS | 5323-87-5 |
|---|---|
| Molecular Formula | C8H12O2 |
| Molecular Weight (g/mol) | 140.18 |
| MDL Number | MFCD00001580 |
| InChI Key | JWCFJPLIRVYENQ-UHFFFAOYSA-N |
| Synonym | 3-ethoxy-2-cyclohexen-1-one, 3-ethoxycyclohex-2-enone, 2-cyclohexen-1-one, 3-ethoxy, 3-ethoxy-2-cyclohexenone, 3-ethoxy-2-cyclohexene-1-one, 3-ethoxycyclohex-2-ene-1-one, 1-ethoxycyclohexene-3-one, acmc-209l4y, 3-ethoxy-2-cyclohexen-one, 3-ethoxy-cyclohex-2-enone |
| PubChem CID | 79216 |
| IUPAC Name | 3-ethoxycyclohex-2-en-1-one |
| SMILES | CCOC1=CC(=O)CCC1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1422409
|
Thermo Scientific Chemicals
A1422409 |
10 g |
Each for $87.90
|
|
|||||
|
AAA1422418
|
Thermo Scientific Chemicals
A1422418 |
50 g |
Each for $293.04
|
|
|||||
|
AAA1422430
|
Thermo Scientific Chemicals
A1422430 |
250 g |
Each for $1,126.26
|
|
|||||
Description
3-Ethoxy-2-cyclohexen-1-one is a useful intermediate for the preparation of 3-alkyl- or aryl-2-cyclohexen-1-ones, by addition of organometallic reagents and subsequent hydrolysis. It helps in kinetic enolate formation (LDA) and alkylation occurs at the 6-position, the Stork-Danheiser kinetic alkylation procedure, which has found extensive use in alicyclic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Ethoxy-2-cyclohexen-1-one is a useful intermediate for the preparation of 3-alkyl- or aryl-2-cyclohexen-1-ones, by addition of organometallic reagents and subsequent hydrolysis. It helps in kinetic enolate formation (LDA) and alkylation occurs at the 6-position, the Stork-Danheiser kinetic alkylation procedure, which has found extensive use in alicyclic synthesis.
Solubility
Slightly soluble in water.
Notes
Store at 4°C. Protect from heat. Store away from oxidizing agents.
Chemical Identifiers
| 5323-87-5 | |
| 140.18 | |
| JWCFJPLIRVYENQ-UHFFFAOYSA-N | |
| 79216 | |
| CCOC1=CC(=O)CCC1 |
| C8H12O2 | |
| MFCD00001580 | |
| 3-ethoxy-2-cyclohexen-1-one, 3-ethoxycyclohex-2-enone, 2-cyclohexen-1-one, 3-ethoxy, 3-ethoxy-2-cyclohexenone, 3-ethoxy-2-cyclohexene-1-one, 3-ethoxycyclohex-2-ene-1-one, 1-ethoxycyclohexene-3-one, acmc-209l4y, 3-ethoxy-2-cyclohexen-one, 3-ethoxy-cyclohex-2-enone | |
| 3-ethoxycyclohex-2-en-1-one |
Specifications
| 5323-87-5 | |
| 76°C to 78°C (1 mmHg) | |
| C8H12O2 | |
| MFCD00001580 | |
| 2042049 | |
| Slightly soluble in water. | |
| CCOC1=CC(=O)CCC1 | |
| 140.18 | |
| 140.18 | |
| 3-Ethoxy-2-cyclohexen-1-one |
| 1.04 | |
| 107°C (224°F) | |
| 1.506 | |
| 10 g | |
| 3-ethoxy-2-cyclohexen-1-one, 3-ethoxycyclohex-2-enone, 2-cyclohexen-1-one, 3-ethoxy, 3-ethoxy-2-cyclohexenone, 3-ethoxy-2-cyclohexene-1-one, 3-ethoxycyclohex-2-ene-1-one, 1-ethoxycyclohexene-3-one, acmc-209l4y, 3-ethoxy-2-cyclohexen-one, 3-ethoxy-cyclohex-2-enone | |
| JWCFJPLIRVYENQ-UHFFFAOYSA-N | |
| 3-ethoxycyclohex-2-en-1-one | |
| 79216 | |
| 99% |
Safety and Handling
EINECSNumber : 226-190-7
TSCA : No
Recommended Storage : Store at -20°C
RUO – Research Use Only