missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(Ethoxycarbonyl)phenyl isocyanate, 97%, Thermo Scientific Chemicals

$69.22 - $553.33

Chemical Identifiers

CAS 67531-68-4
Molecular Formula C10H9NO3
Molecular Weight (g/mol) 191.19
MDL Number MFCD00013865
InChI Key UPXGGIRFDZENRD-UHFFFAOYSA-N
Synonym 3-ethoxycarbonyl phenyl isocyanate, 3-carbethoxyphenyl isocyanate, 3-ethoxycarbonylphenyl isocyanate, 3-ethoxycarbonyl phenylisocyanate, 3-isocyanatobenzoic acid ethyl ester, 3-carboethoxyphenyl isocyanate, #, ethyl 3-isocyanatobenzoate, 3-ethoxycarbonyl phenylisothiocyanate, 3-isocyanato-benzoic acid ethyl ester
PubChem CID 589806
IUPAC Name ethyl 3-isocyanatobenzoate
SMILES CCOC(=O)C1=CC(=CC=C1)N=C=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0949503
View Documents
Thermo Scientific Chemicals
L0949503
1 g
Each for $69.22
 
AAL0949506
View Documents
Thermo Scientific Chemicals
L0949506
5 g
Each for $186.27
 
AAL0949514
View Documents
Thermo Scientific Chemicals
L0949514
25 g
Each for $553.33
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

67531-68-4
191.19
UPXGGIRFDZENRD-UHFFFAOYSA-N
589806
CCOC(=O)C1=CC(=CC=C1)N=C=O
C10H9NO3
MFCD00013865
3-ethoxycarbonyl phenyl isocyanate, 3-carbethoxyphenyl isocyanate, 3-ethoxycarbonylphenyl isocyanate, 3-ethoxycarbonyl phenylisocyanate, 3-isocyanatobenzoic acid ethyl ester, 3-carboethoxyphenyl isocyanate, #, ethyl 3-isocyanatobenzoate, 3-ethoxycarbonyl phenylisothiocyanate, 3-isocyanato-benzoic acid ethyl ester
ethyl 3-isocyanatobenzoate

Specifications

67531-68-4
213°C
C10H9NO3
MFCD00013865
UN2206
Moisture sensitive
UPXGGIRFDZENRD-UHFFFAOYSA-N
ethyl 3-isocyanatobenzoate
589806
97%
1.154
>110°C (230°F)
1.531
1 g
7636626
3-ethoxycarbonyl phenyl isocyanate, 3-carbethoxyphenyl isocyanate, 3-ethoxycarbonylphenyl isocyanate, 3-ethoxycarbonyl phenylisocyanate, 3-isocyanatobenzoic acid ethyl ester, 3-carboethoxyphenyl isocyanate, #, ethyl 3-isocyanatobenzoate, 3-ethoxycarbonyl phenylisothiocyanate, 3-isocyanato-benzoic acid ethyl ester
CCOC(=O)C1=CC(=CC=C1)N=C=O
191.19
191.19
3-(Ethoxycarbonyl)phenyl isocyanate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P311-P330-P333+P313-P362-P501c

H302-H315-H317-H319-H331-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only