Learn More
3-(Ethoxycarbonyl)phenylzinc iodide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H58105AD
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
3-(Ethoxycarbonyl)phenylzinc iodide | |
282727-18-8 | |
C9H9IO2Zn | |
50 mL | |
Air sensitive | |
KNTSJWIWLNBYJL-UHFFFAOYSA-M | |
ethyl benzoate;iodozinc(1+) | |
6095944 |
Packaged under Argon in resealable ChemSeal™ bottles | |
0.991 | |
MFCD00671981 | |
UN3399 | |
zinc 2+ ion 3-ethoxycarbonyl benzen-1-ide iodide | |
CCOC(=O)C1=C[C-]=CC=C1.[Zn+]I | |
341.453 | |
341.45 |
Chemical Identifiers
282727-18-8 | |
341.453 | |
KNTSJWIWLNBYJL-UHFFFAOYSA-M | |
6095944 | |
CCOC(=O)C1=C[C-]=CC=C1.[Zn+]I |
C9H9IO2Zn | |
MFCD00671981 | |
zinc 2+ ion 3-ethoxycarbonyl benzen-1-ide iodide | |
ethyl benzoate;iodozinc(1+) |
Safety and Handling
GHS H Statement
H225-H261-H314-H351-H335
Highly flammable liquid and vapor.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
May cause respiratory irritation.
P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H225-H302-H314-H335-H336-H351
DOTInformation : Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE
TSCA : No
Recommended Storage : Keep cold; Store under Argon