missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Ethylbiphenyl 98.0+%, TCI America™
Supplier: TCI America E056825G
Specifications
| 3-Ethylbiphenyl | |
| -27°C | |
| 284°C | |
| MFCD00796638 | |
| HUXKTWJQSHBZIV-UHFFFAOYSA-N | |
| 1-ethyl-3-phenylbenzene | |
| 79752 | |
| ≥98.0% (GC) |
| 5668-93-9 | |
| Colorless | |
| C14H14 | |
| 25 g | |
| CCC1=CC=CC(=C1)C2=CC=CC=C2 | |
| 182.266 | |
| 182.27 | |
| Liquid |
Chemical Identifiers
| 5668-93-9 | |
| 182.266 | |
| HUXKTWJQSHBZIV-UHFFFAOYSA-N | |
| 1-ethyl-3-phenylbenzene |
| C14H14 | |
| MFCD00796638 | |
| 79752 | |
| CCC1=CC=CC(=C1)C2=CC=CC=C2 |
Safety and Handling
EINECSNumber : (4)-0015
TSCA : No