Learn More
3-Ethyltoluene, 97%, Thermo Scientific Chemicals
$115.22 - $398.86
Chemical Identifiers
CAS | 620-14-4 |
---|---|
Molecular Formula | C9H12 |
Molecular Weight (g/mol) | 120.20 |
MDL Number | MFCD00009259 |
InChI Key | ZLCSFXXPPANWQY-UHFFFAOYSA-N |
Synonym | 3-ethyltoluene, m-ethyltoluene, 1-methyl-3-ethylbenzene, benzene, 1-ethyl-3-methyl, m-ethylmethylbenzene, m-methylethylbenzene, toluene, m-ethyl, 3-methylethylbenzene, 1-ethyl-3-methyl-benzene, unii-737ptd7o7e |
PubChem CID | 12100 |
ChEBI | CHEBI:77512 |
IUPAC Name | 1-ethyl-3-methylbenzene |
SMILES | CCC1=CC=CC(C)=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1878806
|
Thermo Scientific Chemicals
L1878806 |
5 g |
Each for $115.22
|
|
|||||
AAL1878814
|
Thermo Scientific Chemicals
L1878814 |
25 g |
Each for $398.86
|
|
|||||
Description
It is used in the formation of 3-ethylsalicylic acid from 3-Ethyltoluene by Pseudomonas ovalis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used in the formation of 3-ethylsalicylic acid from 3-Ethyltoluene by Pseudomonas ovalis.
Solubility
Not miscible or difficult to mix in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep ignition sources away. Protect from electrostatic charges.
Chemical Identifiers
620-14-4 | |
120.20 | |
ZLCSFXXPPANWQY-UHFFFAOYSA-N | |
12100 | |
1-ethyl-3-methylbenzene |
C9H12 | |
MFCD00009259 | |
3-ethyltoluene, m-ethyltoluene, 1-methyl-3-ethylbenzene, benzene, 1-ethyl-3-methyl, m-ethylmethylbenzene, m-methylethylbenzene, toluene, m-ethyl, 3-methylethylbenzene, 1-ethyl-3-methyl-benzene, unii-737ptd7o7e | |
CHEBI:77512 | |
CCC1=CC=CC(C)=C1 |
Specifications
620-14-4 | |
0.865 | |
38°C (100°F) | |
1.496 | |
5 g | |
1850821 | |
Not miscible or difficult to mix in water. | |
CCC1=CC=CC(C)=C1 | |
120.20 | |
CHEBI:77512 | |
≥98% |
-96°C | |
158°C to 159°C | |
C9H12 | |
MFCD00009259 | |
UN3295 | |
3-ethyltoluene, m-ethyltoluene, 1-methyl-3-ethylbenzene, benzene, 1-ethyl-3-methyl, m-ethylmethylbenzene, m-methylethylbenzene, toluene, m-ethyl, 3-methylethylbenzene, 1-ethyl-3-methyl-benzene, unii-737ptd7o7e | |
ZLCSFXXPPANWQY-UHFFFAOYSA-N | |
1-ethyl-3-methylbenzene | |
12100 | |
120.2 | |
3-Ethyltoluene |
Safety and Handling
GHS H Statement
H226
Flammable liquid and vapor.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c
H226-H304-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
EINECSNumber : 210-626-8
RTECSNumber : DA0712000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only