missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Ethynylaniline 98.0+%, TCI America™
Supplier: TCI America A112225G
Specifications
3-Ethynylaniline | |
Brown | |
C8H7N | |
25 g | |
NNKQLUVBPJEUOR-UHFFFAOYSA-N | |
3-ethynylaniline | |
104682 | |
≥98.0% (GC) |
54060-30-9 | |
120°C | |
MFCD00014779 | |
3-aminophenylacetylene, benzenamine, 3-ethynyl, m-aminophenylacetylene, 3-ethynylbenzenamine, 3-ethynylphenylamine, 3-ethynylaniline, m-aminophenyl acetylene, 3-aminophenyl acetylene, m-ethynylaniline, 3-ethynyl-aniline | |
NC1=CC=CC(=C1)C#C | |
117.15 | |
117.15 | |
Liquid |
Safety and Handling
TSCA : Yes