missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Fluoro-1,2-dihydroxybenzene, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 264790010

 View more versions of this product

Catalog No. AC264790010


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-Fluoro-1, 2-dihydroxybenzene
363-52-0
100.0
Beige to Brown-Gray
98+%
C6H5FO2
1 g
Solubility in water: >350g/L (20°C). Other solubilities: soluble in alcohols,ketones,esters
OC1=CC=CC(F)=C1O
128.10
CHEBI:39876
98+%
98+%
98.0
71°C to 74°C
Authentic
Glass bottle
MFCD00042582
3-fluorocatechol, pyrocatechol, 3-fluoro, 1,2-benzenediol, 3-fluoro, 1,2-dihydroxy-3-fluorobenzene, 3-fluoro-1,2-dihydroxybenzene, 3-fluoro-1,2-benzenediol, 1-fluoro-2,3-dihydroxybenzene, 3fa, pubchem4142
DXOSJQLIRGXWCF-UHFFFAOYSA-N
3-fluorobenzene-1,2-diol
67764
128.1
Crystalline Powder

Chemical Identifiers

363-52-0
128.10
DXOSJQLIRGXWCF-UHFFFAOYSA-N
67764
3-fluorobenzene-1,2-diol
C6H5FO2
MFCD00042582
3-fluorocatechol, pyrocatechol, 3-fluoro, 1,2-benzenediol, 3-fluoro, 1,2-dihydroxy-3-fluorobenzene, 3-fluoro-1,2-dihydroxybenzene, 3-fluoro-1,2-benzenediol, 1-fluoro-2,3-dihydroxybenzene, 3fa, pubchem4142
CHEBI:39876
OC1=CC=CC(F)=C1O
Videos
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only