missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-2-methylaniline 98.0+%, TCI America™

$91.33 - $275.87

Chemical Identifiers

CAS 443-86-7
Molecular Formula C7H8FN
Molecular Weight (g/mol) 125.15
MDL Number MFCD00007760
InChI Key SLDLVGFPFFLYBM-UHFFFAOYSA-N
Synonym 3-fluoro-o-toluidine, 2-fluoro-6-aminotoluene, 2-amino-6-fluorotoluene, 6-amino-2-fluorotoluene, 3-fluoro-2-methyl-aniline, benzenamine, 3-fluoro-2-methyl, 3-fluoro-2-methylphenylamine, 2-methyl-3-fluoroaniline, hydrochloride, 3-fluor-2-methylanilin
PubChem CID 285553
IUPAC Name 3-fluoro-2-methylaniline
SMILES CC1=C(N)C=CC=C1F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
F05265G
View Documents
TCI America
F05265G
5 g
Each for $91.33
Only null left
 
F052625G
View Documents
TCI America
F052625G
25 g
Each for $275.87
Only null left
 
Description

Description

Specifications

Chemical Identifiers

443-86-7
125.15
SLDLVGFPFFLYBM-UHFFFAOYSA-N
285553
CC1=C(N)C=CC=C1F
C7H8FN
MFCD00007760
3-fluoro-o-toluidine, 2-fluoro-6-aminotoluene, 2-amino-6-fluorotoluene, 6-amino-2-fluorotoluene, 3-fluoro-2-methyl-aniline, benzenamine, 3-fluoro-2-methyl, 3-fluoro-2-methylphenylamine, 2-methyl-3-fluoroaniline, hydrochloride, 3-fluor-2-methylanilin
3-fluoro-2-methylaniline

Specifications

443-86-7
C7H8FN
5 g
SLDLVGFPFFLYBM-UHFFFAOYSA-N
3-fluoro-2-methylaniline
285553
≥98.0% (GC,T)
3-Fluoro-2-methylaniline
205°C
MFCD00007760
3-fluoro-o-toluidine, 2-fluoro-6-aminotoluene, 2-amino-6-fluorotoluene, 6-amino-2-fluorotoluene, 3-fluoro-2-methyl-aniline, benzenamine, 3-fluoro-2-methyl, 3-fluoro-2-methylphenylamine, 2-methyl-3-fluoroaniline, hydrochloride, 3-fluor-2-methylanilin
CC1=C(N)C=CC=C1F
125.15
125.15
Liquid
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'