missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Fluoro-2-nitrophenol, 97%, Thermo Scientific Chemicals

$68.70 - $259.41

Chemical Identifiers

CAS 385-01-3
Molecular Formula C6H4FNO3
Molecular Weight (g/mol) 157.1
MDL Number MFCD07368753
InChI Key GAWNBKUTBVLIPL-UHFFFAOYSA-N
Synonym 3-fluoro-2-nitro-phenol, 2-nitro-3-fluorophenol, phenol, 3-fluoro-2-nitro, 3-fluoronitro-phenol, pubchem4124, acmc-209j0i, ksc497m1h
PubChem CID 2782777
IUPAC Name 3-fluoro-2-nitrophenol
SMILES C1=CC(=C(C(=C1)F)[N+](=O)[O-])O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6167003
View Documents
Thermo Scientific Chemicals
H6167003
1 g
Each for $68.70
 
AAH6167006
View Documents
Thermo Scientific Chemicals
H6167006
5 g
Each for $259.41
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

385-01-3
157.1
GAWNBKUTBVLIPL-UHFFFAOYSA-N
2782777
C1=CC(=C(C(=C1)F)[N+](=O)[O-])O
C6H4FNO3
MFCD07368753
3-fluoro-2-nitro-phenol, 2-nitro-3-fluorophenol, phenol, 3-fluoro-2-nitro, 3-fluoronitro-phenol, pubchem4124, acmc-209j0i, ksc497m1h
3-fluoro-2-nitrophenol

Specifications

385-01-3
MFCD07368753
3-fluoro-2-nitro-phenol, 2-nitro-3-fluorophenol, phenol, 3-fluoro-2-nitro, 3-fluoronitro-phenol, pubchem4124, acmc-209j0i, ksc497m1h
C1=CC(=C(C(=C1)F)[N+](=O)[O-])O
157.1
157.1
Solid
C6H4FNO3
1 g
GAWNBKUTBVLIPL-UHFFFAOYSA-N
3-fluoro-2-nitrophenol
2782777
97%
3-Fluoro-2-nitrophenol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only