Learn More
3-Fluoro-2-nitrotoluene, 96%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H6357403
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
3-Fluoro-2-nitrotoluene | |
17°C to 18°C | |
C7H6FNO2 | |
1 g | |
3-fluoro-2-nitrotoluene, 1-fluoro-3-methyl-2-nitro-benzene, 2-fluoro-6-methylnitrobenzene, benzene, 1-fluoro-3-methyl-2-nitro, fluoronitrotoluene, fluor-nitro-toluole, pubchem18581, 2-nitro-3-fluorotoluene, ksc497k3l, 3-methyl-2-nitrofluorobenzene | |
CC1=C(C(=CC=C1)F)[N+](=O)[O-] | |
155.128 | |
155.13 | |
Low Melting Solid |
3013-27-2 | |
White | |
MFCD08458093 | |
UN2811 | |
JSXAJLMUBSEJFF-UHFFFAOYSA-N | |
1-fluoro-3-methyl-2-nitrobenzene | |
19773803 | |
96% |
Chemical Identifiers
3013-27-2 | |
155.128 | |
JSXAJLMUBSEJFF-UHFFFAOYSA-N | |
19773803 | |
CC1=C(C(=CC=C1)F)[N+](=O)[O-] |
C7H6FNO2 | |
MFCD08458093 | |
3-fluoro-2-nitrotoluene, 1-fluoro-3-methyl-2-nitro-benzene, 2-fluoro-6-methylnitrobenzene, benzene, 1-fluoro-3-methyl-2-nitro, fluoronitrotoluene, fluor-nitro-toluole, pubchem18581, 2-nitro-3-fluorotoluene, ksc497k3l, 3-methyl-2-nitrofluorobenzene | |
1-fluoro-3-methyl-2-nitrobenzene |
Safety and Handling
GHS H Statement
H302-H332-H315-H319-H335
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H332-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold