missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Fluoro-2-(trifluoromethyl)aniline, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals H3268403

 View more versions of this product

Catalog No. AAH3268403


Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

3-Fluoro-2-(trifluoromethyl)aniline
C7H5F4N
MFCD06658260
UN2810
SJMDWKFHBPDEQT-UHFFFAOYSA-N
3-fluoro-2-(trifluoromethyl)aniline
15614240
97%
123973-22-8
1.4695
1 g
3-fluoro-2-trifluoromethyl aniline, 2-amino-6-fluorobenzotrifluoride, benzenamine, 3-fluoro-2-trifluoromethyl, 3-fluoro-2-trifluoromethyl phenylamine, benzenamine,3-fluoro-2-trifluoromethyl, 2-amino-6-fluoro-trifluoromethylbenzene, 3-fluoro-2-trifluoromethyl benzenamine, pubchem4833, 2-trifluoromethyl-3-fluoroaniline, 3-fluoranyl-2-trifluoromethyl aniline
NC1=C(C(F)=CC=C1)C(F)(F)F
179.12
179.12

Chemical Identifiers

123973-22-8
179.12
SJMDWKFHBPDEQT-UHFFFAOYSA-N
3-fluoro-2-(trifluoromethyl)aniline
C7H5F4N
MFCD06658260
15614240
NC1=C(C(F)=CC=C1)C(F)(F)F
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H311-H227-H302-H332-H315-H319
Toxic in contact with skin.
Combustible liquid.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.

P210-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P361-P363-P370+P378q-P501c

H227-H302+H332-H311-H315-H319

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions