missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-hydroxybenzaldehyde, 98%

CAS: 405-05-0 | C7H5FO2 | 140.113 g/mol

Supplier:  Thermo Scientific Chemicals L1932506

 View more versions of this product

Catalog No. AAL1932506


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-Fluoro-4-hydroxybenzaldehyde
121°C to 124°C
MFCD00016626
3234259
3-fluoro-4-hydroxy-benzaldehyde, benzaldehyde, 3-fluoro-4-hydroxy, 4-hydroxy-3-fluorobenzaldehyde, pubchem1443, acmc-1acq8, ksc497m8t, 3-fluoro-4 hydroxybenzaldehyde, 4-hydroxy-3-fluoro-benzaldehyde, #, 3-fluoranyl-4-oxidanyl-benzaldehyde
C1=CC(=C(C=C1C=O)F)O
140.113
140.11
405-05-0
C7H5FO2
5 g
Air Sensitive
QSBHJTCAPWOIIE-UHFFFAOYSA-N
3-fluoro-4-hydroxybenzaldehyde
587250
98%

Chemical Identifiers

405-05-0
140.113
QSBHJTCAPWOIIE-UHFFFAOYSA-N
587250
C1=CC(=C(C=C1C=O)F)O
C7H5FO2
MFCD00016626
3-fluoro-4-hydroxy-benzaldehyde, benzaldehyde, 3-fluoro-4-hydroxy, 4-hydroxy-3-fluorobenzaldehyde, pubchem1443, acmc-1acq8, ksc497m8t, 3-fluoro-4 hydroxybenzaldehyde, 4-hydroxy-3-fluoro-benzaldehyde, #, 3-fluoranyl-4-oxidanyl-benzaldehyde
3-fluoro-4-hydroxybenzaldehyde
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only